Skip to Content
Merck
All Photos(1)

Key Documents

EHU129271

Sigma-Aldrich

MISSION® esiRNA

targeting human DDX5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAGCTTGCTGAAGATTTCCTGAAAGACTATATTCATATAAACATTGGTGCACTTGAACTGAGTGCAAACCACAACATTCTTCAGATTGTGGATGTGTGTCATGACGTAGAAAAGGATGAAAAACTTATTCGTCTAATGGAAGAGATCATGAGTGAGAAGGAGAATAAAACCATTGTTTTTGTGGAAACCAAAAGAAGATGTGATGAGCTTACCAGAAAAATGAGGAGAGATGGGTGGCCTGCCATGGGTATCCATGGTGACAAGAGTCAACAAGAGCGTGACTGGGTTCTAAATGAATTCAAACATGGAAAAGCTCCTATTCTGATTGCTACAGATGTGGCCTCCAGAGGGCTAGATGTGGAAGATGTGAAATTTGTCATCAATTATGACTACCCTAACTCCTCAGAGGATTATATTCATCGAATTGGAAGAACTGCTCGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nazia Abbasi et al.
Life science alliance, 3(10) (2020-08-21)
Tumorigenesis in different segments of the intestinal tract involves tissue-specific oncogenic drivers. In the colon, complement component 3 (C3) activation is a major contributor to inflammation and malignancies. By contrast, tumorigenesis in the small intestine involves fatty acid-binding protein 1
Hao Zhang et al.
Hepatology (Baltimore, Md.), 69(3), 1046-1063 (2018-10-04)
In hepatocellular carcinoma (HCC), dysregulated expression of DDX5 (DEAD box protein 5) and impaired autophagy have been reported separately. However, the relationship between them has not been explored. Here we present evidence to show that, by interacting with autophagic receptor
Saravana Kumar Kailasam Mani et al.
Theranostics, 10(24), 10957-10972 (2020-10-13)
Rationale: RNA helicase DDX5 is downregulated during hepatitis B virus (HBV) replication, and poor prognosis HBV-related hepatocellular carcinoma (HCC). The aim of this study is to determine the mechanism and significance of DDX5 downregulation for HBV-driven HCC, and identify biologics
Yeon J Lee et al.
Genes & development, 32(15-16), 1060-1074 (2018-07-26)
Alternative premessenger RNA (pre-mRNA) splicing is a post-transcriptional mechanism for controlling gene expression. Splicing patterns are determined by both RNA-binding proteins and nuclear pre-mRNA structure. Here, we analyzed pre-mRNA splicing patterns, RNA-binding sites, and RNA structures near these binding sites
Zheng Xing et al.
RNA (New York, N.Y.), 23(7), 1125-1138 (2017-04-16)
DEAD-box proteins are a class of nonprocessive RNA helicases that dynamically modulate the structure of RNA and ribonucleoprotein complexes (RNPs). However, the precise roles of individual members are not well understood. Work from our laboratory revealed that the DEAD-box protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service