Skip to Content
Merck
All Photos(1)

Key Documents

EHU125021

Sigma-Aldrich

MISSION® esiRNA

targeting human GYPC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACGGAGGTGGGAGAAAATCTGGGCACATGGGGCCCCCTGGGCAGTGCAGGACAACATCAGCTCACTGGCAGGAAAGTCCTTGTTGAGGGTGAGGGGGTGCTGGGGTACCCGGGGGCTGGGGAAGCAAGGAAATAAGTCATCTGTATGCTGACTGGGGATAATGGCATCAAATGTCAGTCCTTGACATTTGGGGGGAACAGCAGGTGCCAGAGCTAAAAGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Liu et al.
Hepatology (Baltimore, Md.), 69(4), 1535-1548 (2018-12-07)
Endocannabinoids promote energy conservation in obesity, whereas cannabinoid-1 receptor (CB1 R) blockade reverses body weight gain and insulin resistance and increases energy expenditure. Here we investigated the molecular mechanisms of the catabolic effects of CB1 R blockade in the liver.
Weiwei Cheng et al.
Neuron, 104(5), 885-898 (2019-10-08)
Hexanucleotide GGGGCC repeat expansion in C9ORF72 is the most prevalent genetic cause of amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD). One pathogenic mechanism is the aberrant accumulation of dipeptide repeat (DPR) proteins produced by the unconventional translation of expanded RNA repeats.
Charles A Berdan et al.
Cell chemical biology, 26(7), 1027-1035 (2019-05-14)
Parthenolide, a natural product from the feverfew plant and member of the large family of sesquiterpene lactones, exerts multiple biological and therapeutic activities including anti-inflammatory and anti-cancer effects. Here, we further study the parthenolide mechanism of action using activity-based protein
Florian Beaumatin et al.
Molecular cell, 76(1), 163-176 (2019-09-08)
Sensing nutrient availability is essential for appropriate cellular growth, and mTORC1 is a major regulator of this process. Mechanisms causing mTORC1 activation are, however, complex and diverse. We report here an additional important step in the activation of mTORC1, which
Arif Yurdagul et al.
Cell metabolism, 31(3), 518-533 (2020-02-01)
Continual efferocytic clearance of apoptotic cells (ACs) by macrophages prevents necrosis and promotes injury resolution. How continual efferocytosis is promoted is not clear. Here, we show that the process is optimized by linking the metabolism of engulfed cargo from initial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service