Skip to Content
Merck
All Photos(1)

Key Documents

EHU120781

Sigma-Aldrich

MISSION® esiRNA

targeting human CNR2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCTGGGTGACAGAGATAGCCAATGGCTCCAAGGATGGCTTGGATTCCAACCCTATGAAGGATTACATGATCCTGAGTGGTCCCCAGAAGACAGCTGTTGCTGTGTTGTGCACTCTTCTGGGCCTGCTAAGTGCCCTGGAGAACGTGGCTGTGCTCTATCTGATCCTGTCCTCCCACCAACTCCGCCGGAAGCCCTCATACCTGTTCATTGGCAGCTTGGCTGGGGCTGACTTCCTGGCCAGTGTGGTCTTTGCATGCAGCTTTGTGAATTTCCATGTTTTCCATGGTGTGGATTCCAAGGCTGTCTTCCTGCTGAAGATTGGCAGCGTGACTATGACCTTCACAGCCTCTGTGGGTAGCCTCCTGCTGACCGCCATTGACCGATACCTCTGCCTGCGCTATCCACCTTCCTACAAAGCTCTGCTCACCCGTGGAAGGGCACTGGTGACCCTGGGCATCATGTGGGTCCTCTCAGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabrina Fechtner et al.
Clinical and experimental rheumatology, 37(6), 1026-1035 (2019-04-04)
Recent studies showed that the expression of cannabinoid receptor 2 (CB2), not CB1, is upregulated at both the mRNA and protein levels in rheumatoid arthritis synovial fibroblasts (RASFs), however, little is known about its endogenous role in pro-inflammatory cytokine signalling
Young Sun Hwang et al.
Chemico-biological interactions, 273, 107-114 (2017-06-12)
Melanogenesis plays a critical role in the protection of skin against external stresses such as ultraviolet irradiation and oxidative stressors. This study was aimed to investigate the effects of cannabidiol on melanogenesis and its mechanisms of action in human epidermal
Lei Tian et al.
Frontiers in immunology, 8, 1214-1214 (2017-10-17)
Macrophage M1/M2 polarization mediates tissue damage and inflammatory responses. Cannabinoid receptor (CB) 1 participated in liver fibrogenesis by affecting bone marrow (BM)-derived monocytes/macrophages (BMMs) activation. However, the knowledge of whether CB1 is involved in the polarization of BMMs remains limited.
Chao Liu et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 26(11), 2693-2703 (2020-01-15)
Human papillomavirus (HPV)-related head and neck squamous cell carcinoma (HNSCC) is associated with daily marijuana use and is also increasing in parallel with increased marijuana use in the United States. Our study is designed to define the interaction between cannabinoids
Sabrina Fechtner et al.
Frontiers in immunology, 10, 1027-1027 (2019-05-30)
Management of pain in the treatment of rheumatoid arthritis (RA) is a priority that is not fully addressed by the conventional therapies. In the present study, we evaluated the efficacy of cannabinoid receptor 2 (CB2) agonist JWH-015 using RA synovial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service