Skip to Content
Merck
All Photos(1)

Key Documents

EHU105821

Sigma-Aldrich

MISSION® esiRNA

targeting human MST1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCAGTAGCCAAGATGGTGTGTGGGCCCTCAGGCTCCCAGCTTGTCCTGCTCAAGCTGGAGAGATCTGTGACCCTGAACCAGCGTGTGGCCCTGATCTGCCTGCCCCCTGAATGGTATGTGGTGCCTCCAGGGACCAAGTGTGAGATTGCAGGCTGGGGTGAGACCAAAGGTACGGGTAATGACACAGTCCTAAATGTGGCCTTGCTGAATGTCATCTCCAACCAGGAGTGTAACATCAAGCACCGAGGACGTGTGCGGGAGAGTGAGATGTGCACTGAGGGACTGTTGGCCCCTGTGGGGGCCTGTGAGGGTGACTACGGGGGCCCACTTGCCTGCTTTACCCACAACTGCTGGGTCCTGGAAGGAATTATAATCCCCAACCGAGTATGCGCAAGGTCCCGCTGGCCAGCTGTCTTCACGCGTGTCTCTGTGT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trang Nguyen-Vu et al.
Breast cancer research : BCR, 15(3), R51-R51 (2013-07-03)
Liver × receptors (LXRs) are members of the nuclear receptor family of ligand-dependent transcription factors and have established functions as regulators of cholesterol, glucose, and fatty acid metabolism and inflammatory responses. Published reports of anti-proliferative effects of synthetic LXR ligands
Li Chen et al.
International journal of clinical and experimental pathology, 8(9), 10545-10554 (2015-12-01)
E2F transcription factors regulate a wide range of biological processes, including cell cycle, apoptosis and DNA damage response. In the present study, we examined whether E2F2 is related to the poor prognosis of NSCLC and its role in progress of
Zheng-Qing Fang et al.
Journal of cellular biochemistry, 119(10), 8317-8324 (2018-06-23)
We intended to evaluate miR-490-5p expression in hepatocellular carcinoma (HCC) tissues and detect the potential targets of miR-490-5p. In vitro experiments were conducted to further investigate the biological function of miR-490-5p on HCC cell metastasis. We investigated the abnormally expressed
Hang Song et al.
Journal of physiology and biochemistry, 72(4), 733-744 (2016-08-16)
Glioblastoma multiforme (GBM), the most common and lethal primary brain tumor in adults characterized by high proliferative ability and mortality rate, contains a small subpopulation of cancer stem-like cells (CSCs), which is responsible for GBM progression and therapeutic resistance. Numerous
Shiguan Wang et al.
Arthritis research & therapy, 20(1), 225-225 (2018-10-06)
Expression of E2F transcription factor 2 (E2F2), a transcription factor related to the cell cycle, is abnormally high in rheumatoid arthritis synovial fibroblasts (RASFs). Deregulated expression of E2F2 leads to abnormal production of proinflammatory cytokines, such as interleukin (IL)-1α, IL-1β

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service