Skip to Content
Merck
All Photos(1)

Key Documents

EHU084671

Sigma-Aldrich

MISSION® esiRNA

targeting human COPS5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGCACTGAAACCCGAGTAAATGCTCAGGCTGCTGCATATGAATACATGGCTGCATACATAGAAAATGCAAAACAGGTTGGCCGCCTTGAAAATGCAATCGGGTGGTATCATAGCCACCCTGGCTATGGCTGCTGGCTTTCTGGGATTGATGTTAGTACTCAGATGCTCAATCAGCAGTTCCAGGAACCATTTGTAGCAGTGGTGATTGATCCAACAAGAACAATATCCGCAGGGAAAGTGAATCTTGGCGCCTTTAGGACATACCCAAAGGGCTACAAACCTCCTGATGAAGGACCTTCTGAGTACCAGACTATTCCACTTAATAAAATAGAAGATTTTGGTGTACACTGCAAACAATATTATGCCTTAGAAGTCTCATATTTCAAATCCTCTTTGGATCGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anja Schwarz et al.
Journal of biomedical science, 24(1), 12-12 (2017-02-09)
Oxidized low-density lipoprotein (oxLDL) mediates the transformation of macrophages (MΦ) to cholesterol-rich foam cells and the release of pro-inflammatory cytokines during atherogenesis. JAB1 (Jun activation domain binding protein-1) is present in all stages of human plaques, involved in the Toll-like
Yang Liu et al.
Acta pharmaceutica Sinica. B, 10(12), 2299-2312 (2020-12-24)
Programmed cell death-1 (PD-1)/programmed cell death ligand-1 (PD-L1) blocking therapy has become a major pillar of cancer immunotherapy. Compared with antibodies targeting, small-molecule checkpoint inhibitors which have favorable pharmacokinetics are urgently needed. Here we identified berberine (BBR), a proven anti-inflammation
S Wang et al.
Oncogene, 35(47), 6096-6108 (2016-05-10)
Radiotherapy is the standard therapy for nasopharyngeal carcinoma (NPC); however, radioresistance can hinder successful treatment. Here we report that microRNA (miR)-24 acts as a tumor suppressor and radiosensitizer in NPC cells and xenografts by targeting Jab1/CSN5. Although accumulating evidence has
Lin Wang et al.
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association, 23(6), 1003-1017 (2020-05-28)
Jab1 has been reported to regulate various proteins in signal transduction pathways and be implicated in carcinogenesis or tumor progression. However, the precise role and molecular mechanism of Jab1 in gastric tumorigenesis have not yet been fully elucidated. Jab1 staining
Sandra Jumpertz et al.
Cellular signalling, 34, 38-46 (2017-02-24)
The COP9 signalosome (CSN) is a multi-protein complex that is highly conserved in eukaryotes. Due to its regulatory impact on processes such as cell cycle, DNA damage response and apoptosis, the CSN is essential for mammalian cells. One of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service