Skip to Content
Merck
All Photos(1)

Key Documents

EHU079921

Sigma-Aldrich

MISSION® esiRNA

targeting human MCL1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTGCTCCCCATTGATTGAAGAGTCACTGTCTGAAAGAAGCAAAGTTCAGTTTCAGCAACAAACAAACTTTGTTTGGGAAGCTATGGAGGAGGACTTTTAGATTTAGTGAAGATGGTAGGGTGGAAAGACTTAATTTCCTTGTTGAGAACAGGAAAGTGGCCAGTAGCCAGGCAAGTCATAGAATTGATTACCCGCCGAATTCATTAATTTACTGTAGTGTTAAGAGAAGCACTAAGAATGCCAGTGACCTGTGTAAAAGTTACAAGTAATAGAACTATGACTGTAAGCCTCAGTACTGTACAAGGGAAGCTTTTCCTCTCTCTAATTAGCTTTCCCAGTATACTTCTTAGAAAGTCCAAGTGTTCAGGACTTTTATACCTGTTATACTTTGGCTTGGTTTCCATG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eimear O' Reilly et al.
Scientific reports, 8(1), 15752-15752 (2018-10-27)
Acute myeloid leukaemia (AML) is an aggressive cancer with 50-75% of patients relapsing even after successful chemotherapy. The role of the bone marrow microenvironment (BMM) in protecting AML cells from chemotherapeutics and causing consequent relapse is increasingly recognised. However the
Yuzu Zhao et al.
Cell death & disease, 8(10), e3133-e3133 (2017-10-27)
Demethylzeylasteral is one of the extracts of Tripterygium wilfordii Hook F, which plays important roles in multiple biological processes such as inflammation inhibition, as well as immunosuppression. However, anti-cancer function and the underlying mechanisms of demethylzeylasteral in melanoma cells remain
Takahito Sugase et al.
Cancer research, 77(24), 6975-6986 (2017-10-19)
STAT3 has been implicated recently in radioresistance in cancer. In this study, we investigated the association between STAT3 and radioresistance in esophageal squamous cell carcinoma (ESCC). Strong expression of activated phospho-STAT3 (p-STAT3) was observed in 16/22 ESCC patients with preoperative
Yuto Yasuda et al.
Cell death & disease, 11(3), 177-177 (2020-03-11)
There have been few advances in the treatment of small-cell lung cancer (SCLC) because of the lack of targets. MCL1, a member of the anti-apoptotic BCL-2 family, may be a treatment target in several cancers, including SCLC. However, whether the
Weiguo Zhang et al.
Molecular cancer therapeutics, 13(7), 1848-1859 (2014-04-18)
Aberrant activation of multiple signaling pathways is common in acute myelogenous leukemia (AML) cells, which can be linked to a poor prognosis for patients with this disease. Previous research with mTOR or MEK inhibitors revealed cytostatic, rather than cytotoxic, effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service