Skip to Content
Merck
All Photos(1)

Key Documents

EHU078251

Sigma-Aldrich

MISSION® esiRNA

targeting human CTH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GATTCGAAAGCCTTGCTGAGCTTCCGGCAATCATGACTCATGCATCAGTTCTTAAGAATGACAGAGATGTCCTTGGAATTAGTGACACACTGATTCGACTTTCTGTGGGCTTAGAGGATGAGGAAGACCTACTGGAAGATCTAGATCAAGCTTTGAAGGCAGCACACCCTCCAAGTGGAAGTCACAGCTAGTATTCCAGAGCTGCTATTAGAAGCTGCTTCCTGTGAAGATCAAATCTTCCTGAGTAATTAAATGGACCAACAATGAGCCTTTGCAAAATTTTCAAGCGGAAATTTTAAGGCACCTCATTATCTTTCATAACTGTAATTTTCTTAGGGATCATCTCTGTTAAAAAGTTTTCTGTATGTCATGTTATAATTACAGGTCAATTCTGTTAATATCTTTTTGTTAATTTTGCTCTATGTTTGCCTCTGAAGGAGGT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neil Dufton et al.
Scientific reports, 2, 499-499 (2012-07-13)
Hydrogen sulfide is an essential gasotransmitter associated with numerous pathologies. We assert that hydrogen sulfide plays an important role in regulating macrophage function in response to subsequent inflammatory stimuli, promoting clearance of leukocyte infiltrate and reducing TNF-α levels in vivo
Zhongjian Cheng et al.
Circulation, 134(19), 1467-1483 (2016-09-24)
Bone marrow cell (BMC)-based treatment for critical limb ischemia in diabetic patients yielded a modest therapeutic effect resulting from cell dysfunction. Therefore, approaches that improve diabetic stem/progenitor cell functions may provide therapeutic benefits. Here, we tested the hypothesis that restoration
Wen-Long Xue et al.
American journal of physiology. Cell physiology, 318(5), C857-C869 (2020-03-19)
Diabetes (especially Type II) is one of the primary threats to cardiovascular health. Wound healing defects and vascular dysfunction are common in diabetic patients, and the primary cause of deterioration is sustained high plasma glucose. microRNA, a noncoding RNA, has
Daoliang Xu et al.
Pharmacological research, 117, 357-369 (2017-01-15)
It has been suggested that excessive apoptosis in intervertebral disc cells induced by inflammatory cytokines, such as interleukin (IL)-1β, is related to the process of intervertebral disc degeneration (IVDD). Hydrogen sulfide (H
Qian-Rong Qi et al.
Endocrinology, 161(11) (2020-09-29)
Angiogenesis is a physiological process for endometrial regeneration in the menstrual cycle and remodeling during pregnancy. Endogenous hydrogen sulfide (H2S), produced by cystathionine-β synthase (CBS) and cystathionine-γ lyase (CSE), is a potent proangiogenic factor; yet, whether the H2S system is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service