Skip to Content
Merck
All Photos(1)

Key Documents

EHU072181

Sigma-Aldrich

MISSION® esiRNA

targeting human PITX2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTCAAAGGGATGTCCTCAGTGTCTGACATCTTTCACTACAAGTATTTCTAACAGTTGCAAGGACACATACACAAACAAATGTTTGACTGGATATGACATTTTAACATTACTATAAGCTTGTTATTTTTTAAGTTTAGCATTGTTAACATTTAAATGACTGAAAGGATGTATATATATCGAAATGTCAAATTAATTTTATAAAAGCAGTTGTTAGTAATATCACAACAGTGTTTTTAAAGGTTAGGCTTTAAAATAAAGCATGTTATACAGAAGCGATTAGGATTTTTCGCTTGCGAGCAAGGGAGTGTATATACTAAATGCCACACTGTATGTTTCTAACATATTATTATTATTATAAAAAATGTGTGAATATCAGTTTTAGAATAGTTTCTCTGGTGGATGCAATGATG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Hsun Kao et al.
European journal of clinical investigation, 49(10), e13160-e13160 (2019-08-06)
A Pitx2c deficiency increases the risk of atrial fibrillation (AF). Atrial structural remodelling with fibrosis blocks electrical conduction and leads to arrhythmogenesis. A Pitx2c deficiency enhances profibrotic transforming growth factor (TGF)-β expression and calcium dysregulation, suggesting that Pitx2c may play
Estefanía Lozano-Velasco et al.
Cardiovascular research, 109(1), 55-66 (2015-08-06)
Atrial fibrillation (AF) is the most common type of arrhythmia in humans, yet the genetic cause of AF remains elusive. Genome-wide association studies (GWASs) have reported risk variants in four distinct genetic loci, and more recently, a meta-GWAS has further
Wing-Kee Lee et al.
Cancer letters, 449, 237-251 (2019-02-12)
Oncogenic pituitary homeobox 2 (PITX2), a de facto master regulator of developmental organ asymmetry, previously upregulated multidrug resistance (MDR) P-glycoprotein ABCB1 in A498 renal cell carcinoma (RCC) cells. The role of PITX2 isoforms in MDR cancers was investigated. Data mining
Estefanía Lozano-Velasco et al.
Molecular and cellular biology, 35(17), 2892-2909 (2015-06-10)
The acquisition of a proliferating-cell status from a quiescent state as well as the shift between proliferation and differentiation are key developmental steps in skeletal-muscle stem cells (satellite cells) to provide proper muscle regeneration. However, how satellite cell proliferation is
Ryuta Tanimoto et al.
Endocrinology, 156(1), 58-70 (2014-11-05)
The growth factor progranulin is as an important regulator of transformation in several cellular systems. We have previously demonstrated that progranulin acts as an autocrine growth factor and stimulates motility, proliferation, and anchorage-independent growth of castration-resistant prostate cancer cells, supporting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service