Skip to Content
Merck
All Photos(1)

Key Documents

EHU062491

Sigma-Aldrich

MISSION® esiRNA

targeting human FYN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CGCAGATCATGAAGAAGCTGAAGCACGACAAGCTGGTCCAGCTCTATGCAGTGGTGTCTGAGGAGCCCATCTACATCGTCACCGAGTATATGAACAAAGGAAGTTTACTGGATTTCTTAAAAGATGGAGAAGGAAGAGCTCTGAAATTACCAAATCTTGTGGACATGGCAGCACAGGTGGCTGCAGGAATGGCTTACATCGAGCGCATGAATTATATCCATAGAGATCTGCGATCAGCAAACATTCTAGTGGGGAATGGACTCATATGCAAGATTGCTGACTTCGGATTGGCCCGATTGATAGAAGACAATGAGTACACAGCAAGACAAGGTGCAAAGTTCCCCATCAAGTGGACGGCCCCCGAGGCAGCCCTGTACGGGAGGTTCACAATCAAGTCTGACGTGTGGTCTTTTGGAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Zheng et al.
Cancer biotherapy & radiopharmaceuticals, 32(9), 320-326 (2017-11-16)
Tyrosine kinase FYN-a member of the SRC family of kinases-is associated with mediating mitogenic signals and regulating cell cycle entry, growth, and proliferation. It was hypothesized that FYN is upregulated in thyroid carcinoma, which plays a critical role in tumorigenesis.
Michaela Nelson et al.
International journal of cancer, 135(10), 2338-2351 (2014-04-15)
Voltage-gated Na(+) channels (VGSCs) are heteromeric proteins composed of pore-forming α subunits and smaller β subunits. The β subunits are multifunctional channel modulators and are members of the immunoglobulin superfamily of cell adhesion molecules (CAMs). β1, encoded by SCN1B, is
Martin Golkowski et al.
Cell systems, 11(2), 196-207 (2020-08-07)
Hepatocellular carcinoma (HCC) is a complex and deadly disease lacking druggable genetic mutations. The limited efficacy of systemic treatments for advanced HCC implies that predictive biomarkers and drug targets are urgently needed. Most HCC drugs target protein kinases, indicating that
Hadas Grossman et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(8), 3206-3216 (2015-04-30)
Granulosa cells support the developing oocytes and serve as transducers of the ovulatory stimulus induced by LH surge. Fyn kinase is expressed in granulosa cells, though its role in these cells has not been studied. In human embryonic kidney 293T
Nikhil Panicker et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(27), 10058-10077 (2015-07-15)
Sustained neuroinflammation mediated by resident microglia is recognized as a key pathophysiological contributor to many neurodegenerative diseases, including Parkinson's disease (PD), but the key molecular signaling events regulating persistent microglial activation have yet to be clearly defined. In the present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service