Skip to Content
Merck
All Photos(1)

Key Documents

EHU054511

Sigma-Aldrich

MISSION® esiRNA

targeting human IGFBP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AAGCATGGCCTGTACAACCTCAAACAGTGCAAGATGTCTCTGAACGGGCAGCGTGGGGAGTGCTGGTGTGTGAACCCCAACACCGGGAAGCTGATCCAGGGAGCCCCCACCATCCGGGGGGACCCCGAGTGTCATCTCTTCTACAATGAGCAGCAGGAGGCTCGCGGGGTGCACACCCAGCGGATGCAGTAGACCGCAGCCAGCCGGTGCCTGGCGCCCCTGCCCCCCGCCCCTCTCCAAACACCGGCAGAAAACGGAGAGTGCTTGGGTGGTGGGTGCTGGAGGATTTTCCAGTTCTGACACACGTATTTATATTTGGAAAGAGACCAGCACCGAGCTCGGCACCTCCCCGGCCTCTCTCTTCCCAGCTGCAGATGCCACACCTGCTCCTTCTTGCTTT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiromichi Katayama et al.
Scientific reports, 7(1), 12016-12016 (2017-09-22)
Renal cell carcinoma (RCC) is one of the most lethal urologic cancers. About one-third of RCC patients already have distal metastasis at the time of diagnosis. There is growing evidence that Hox antisense intergenic RNA (HOTAIR) plays essential roles in
Z-S Yuan et al.
European review for medical and pharmacological sciences, 21(22), 5072-5080 (2017-12-12)
Gliomas are accompanied with high mortality owning to their invasive peculiarity and vulnerability to drug resistance. miR-21 is a vital oncogenic miRNA that regulates drug resistance of tumor cells. This study aims to elucidate the function of miR-21 in human
S W Yau et al.
International journal of obesity (2005), 39(5), 770-781 (2014-11-06)
IGF-binding protein (IGFBP)-2 is the principal IGFBP produced by white adipocytes during adipogenesis, and circulating levels are reduced in obesity. Overexpression of IGFBP-2 in transgenic mice prevents obesity, but depot-specific effects of IGFBP-2 on adipo/lipogenesis are unknown. The present study
Steven W Yau et al.
Endocrinology, 155(6), 2133-2143 (2014-03-25)
Leptin is produced from white adipose tissue and acts primarily to regulate energy balance. Obesity is associated with leptin resistance and increased circulating levels of leptin. Leptin has recently been shown to influence levels of IGF binding protein-2 (IGFBP-2), a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service