Skip to Content
Merck
All Photos(1)

Key Documents

EHU048191

Sigma-Aldrich

MISSION® esiRNA

targeting human SNAI2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCGGACCCACACATTACCTTGTGTTTGCAAGATCTGCGGCAAGGCGTTTTCCAGACCCTGGTTGCTTCAAGGACACATTAGAACTCACACGGGGGAGAAGCCTTTTTCTTGCCCTCACTGCAACAGAGCATTTGCAGACAGGTCAAATCTGAGGGCTCATCTGCAGACCCATTCTGATGTAAAGAAATACCAGTGCAAAAACTGCTCCAAAACCTTCTCCAGAATGTCTCTCCTGCACAAACATGAGGAATCTGGCTGCTGTGTAGCACACTGAGTGACGCAATCAATGTTTACTCGAACAGAATGCATTTCTTCACTCCGAAGCCAAATGACAAATAAAGTCCAAAGGCATTTTCTCCTGTGCTGACCAACCAAATAATATGTATAGACACACACACATATGCACACACACACACACACACCCACAGAGAGAGAGCTGCAAGAGCATGGAATTCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Hyun Jo et al.
Oncotarget, 8(9), 15441-15452 (2017-01-07)
Retinoblastoma is the most common intraocular cancer in children, affecting 1/20,000 live births. Currently, children with retinoblastoma were treated with chemotherapy using drugs such as carboplatin, vincristine, and etoposide. Unfortunately, if conventional treatment fails, the affected eyes should be removed
Wenyang Li et al.
Genes & development, 34(19-20), 1310-1315 (2020-09-19)
SNAI2/SLUG, a metastasis-promoting transcription factor, is a labile protein that is degraded through the ubiquitin proteasome degradation system. Here, we conducted comprehensive gain- and loss-of-function screens using a human DUB cDNA library of 65 genes and an siRNA library of
Mijung Kwon et al.
Oncotarget, 7(47), 77052-77070 (2016-10-25)
Filamin A interacting protein 1-like (FILIP1L) is an inhibitor of the canonical WNT pathway. WNT/β-catenin signaling and its downstream pathway, epithelial-to-mesenchymal transition (EMT), play a key role in ovarian cancer metastasis and chemoresistance. To study the clinical implications of FILIP1L
Ziliang Wang et al.
Oncotarget, 6(9), 6670-6683 (2015-03-10)
While Aurora-A (Aur A) provokes, BRCA2 restrains primary tumorigenesis, the roles of Aur A and BRCA2 in cancer metastasis remains unclear. Here, we show that the metastatic promoting markers SLUG, FBN1, and MMP2, 9, 13 are either stimulated or suppressed
Adrián Blanco-Gómez et al.
Cancer research, 80(23), 5216-5230 (2020-10-08)
SNAI2 overexpression appears to be associated with poor prognosis in breast cancer, yet it remains unclear in which breast cancer subtypes this occurs. Here we show that excess SNAI2 is associated with a poor prognosis of luminal B HER2+/ERBB2+ breast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service