Skip to Content
Merck
All Photos(1)

Key Documents

EHU047031

Sigma-Aldrich

MISSION® esiRNA

targeting human MMP8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GACAATTCATGGAGCCAGGTTATCCCAAAAGCATATCAGGTGCCTTTCCAGGAATAGAGAGTAAAGTTGATGCAGTTTTCCAGCAAGAACATTTCTTCCATGTCTTCAGTGGACCAAGATATTACGCATTTGATCTTATTGCTCAGAGAGTTACCAGAGTTGCAAGAGGCAATAAATGGCTTAACTGTAGATATGGCTGAAGCAAAATCAAATGTGGCTGTATCCACTTTCAGAATGTTGAAGGGAAGTTCAGCAAGCATTTTCGTTACATTGTGTCCTGCTTATACTTTTCTCAATATTAAGTCATTGTTTCCCATCACTGTATCCATTCTACCTGTCCTCCGTGAAAATATGTTTGGAATATTCCACTATTTGCAGAGGCTTATTCAGTTCTTACACATTCCATCTTACATTAGTGATTCCATCAAAGAGAAGGAAAGTAAGCCTTTTTGTCACC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Schubert-Unkmeir et al.
PLoS pathogens, 6(4), e1000874-e1000874 (2010-05-06)
Disruption of the blood-brain barrier (BBB) is a hallmark event in the pathophysiology of bacterial meningitis. Several inflammatory mediators, such as tumor necrosis factor alpha (TNF-alpha), nitric oxide and matrix metalloproteinases (MMPs), contribute to this disruption. Here we show that
Guanmei Wen et al.
The Journal of biological chemistry, 290(31), 19158-19172 (2015-06-21)
Matrix metalloproteinase-8 (MMP8) has been shown to influence various cellular functions. As monocytes and macrophages (Mφ) express MMP8, we investigated if MMP8 played a role in macrophage differentiation and polarization. MMP8 expression was significantly increased during monocyte differentiation into Mφ.
Muge Sarper et al.
Breast cancer research : BCR, 19(1), 33-33 (2017-03-24)
Normal myoepithelial cells (MECs) play an important tumour-suppressor role in the breast but display an altered phenotype in ductal carcinoma in situ (DCIS), gaining tumour-promoter functions. Matrix metalloproteinase-8 (MMP-8) is expressed by normal MECs but is lost in DCIS. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service