Skip to Content
Merck
All Photos(1)

Key Documents

EHU031671

Sigma-Aldrich

MISSION® esiRNA

targeting human CD81

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCTGTCATGATGTTCGTTGGCTTCCTGGGCTGCTACGGGGCCATCCAGGAATCCCAGTGCCTGCTGGGGACGTTCTTCACCTGCCTGGTCATCCTGTTTGCCTGTGAGGTGGCCGCCGGCATCTGGGGCTTTGTCAACAAGGACCAGATCGCCAAGGATGTGAAGCAGTTCTATGACCAGGCCCTACAGCAGGCCGTGGTGGATGATGACGCCAACAACGCCAAGGCTGTGGTGAAGACCTTCCACGAGACGCTTGACTGCTGTGGCTCCAGCACACTGACTGCTTTGACCACCTCAGTGCTCAAGAACAATTTGTGTCCCTCGGGCAGCAACATCATCAGCAACCTCTTCAAGGAGGACTGCCACCAGAAGATCGATGACCTCTTCTCCGGGAAGCTGTACCTCATCGGCAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Down-regulation of CD81 in human cells producing HCV-E1/E2 retroVLPs.
Ana F Rodrigues et al.
BMC proceedings, 5 Suppl 8, P72-P72 (2012-03-01)
Zhen-yong Keck et al.
PLoS pathogens, 8(4), e1002653-e1002653 (2012-04-19)
The majority of broadly neutralizing antibodies to hepatitis C virus (HCV) are against conformational epitopes on the E2 glycoprotein. Many of them recognize overlapping epitopes in a cluster, designated as antigenic domain B, that contains residues G530 and D535. To
Raphaël Gaudin et al.
PloS one, 8(7), e69450-e69450 (2013-08-08)
During HIV pathogenesis, infected macrophages behave as "viral reservoirs" that accumulate and retain virions within dedicated internal Virus-Containing Compartments (VCCs). The nature of VCCs remains ill characterized and controversial. Using wild-type HIV-1 and a replication-competent HIV-1 carrying GFP internal to
Chien-Te K Tseng et al.
The Journal of experimental medicine, 195(1), 43-49 (2002-01-10)
Infection with hepatitis C virus (HCV) is a leading cause of chronic liver disease worldwide. Little is known about how this virus is able to persist or whether this persistence might be because of its ability to alter the early
Zhihui Chen et al.
PloS one, 6(4), e18933-e18933 (2011-04-29)
HCV infection is often associated with B-cell regulatory control disturbance and delayed appearance of neutralizing antibodies. CD81 is a cellular receptor for HCV and can bind to HCV envelope protein 2 (E2). CD81 also participates to form a B cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service