Skip to Content
Merck
All Photos(1)

Key Documents

EHU025821

Sigma-Aldrich

MISSION® esiRNA

targeting human CDKN1B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGTTAACCCGGGACTTGGAGAAGCACTGCAGAGACATGGAAGAGGCGAGCCAGCGCAAGTGGAATTTCGATTTTCAGAATCACAAACCCCTAGAGGGCAAGTACGAGTGGCAAGAGGTGGAGAAGGGCAGCTTGCCCGAGTTCTACTACAGACCCCCGCGGCCCCCCAAAGGTGCCTGCAAGGTGCCGGCGCAGGAGAGCCAGGATGTCAGCGGGAGCCGCCCGGCGGCGCCTTTAATTGGGGCTCCGGCTAACTCTGAGGACACGCATTTGGTGGACCCAAAGACTGATCCGTCGGACAGCCAGACGGGGTTAGCGGAGCAATGCGCAGGAATAAGGAAGCGACCTGCAACCGACGATTCTTCTACTCAAAACAAAAGAGCCAACAGAACAGAAGAAAATGTTTCAGACGGTTCCCCAAATGCCGGTTCTGTGGAGCAGACGCCCAAGAAGCCTGGCCTCAGAAGACGTCAAACGTAAACAGCTCG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Sun et al.
Oncology letters, 8(4), 1435-1440 (2014-09-10)
Radiotherapy (RT) is a major modality of hepatoma treatment. However, liver tumors often acquire radioresistance, which contributes to RT failure. The exact mechanisms of the radioresistance in hepatoma cells are largely unknown. Glutathione S-transferase M3 (GSTM3) is a phase II
Huimin Wang et al.
Oncology research, 25(9), 1431-1440 (2017-01-22)
Nasopharyngeal carcinoma (NPC) is a common malignancy of the head and neck that arises from the nasopharynx epithelium and is highly invasive. Cyclin-dependent kinase inhibitor 3 (CDKN3) belongs to the dual-specificity protein phosphatase family, which plays a key role in
Galit Ankri-Eliahoo et al.
Journal of vascular surgery, 63(5), 1351-1359 (2015-02-24)
The natural response to arterial occlusive disease is enlargement of collaterals; however, the molecular factors that control collateralization are not well understood. The gene p27(Kip1) (p27) affects human response to arterial injury. Previous studies have shown that overexpression of p27
Alessandra Verdina et al.
Journal of translational medicine, 6, 27-27 (2008-05-24)
Nonsteroidal anti-inflammatory drugs (NSAIDs) have been proposed for prevention and treatment of a variety of human cancers. Piroxicam, in particular, has been recently shown to exert significant anti-tumoral activity in combination with cisplatin (CDDP) on mesothelioma cells. However, the mechanisms
Sabarish Ramachandran et al.
Journal of Korean medical science, 23(4), 667-673 (2008-08-30)
The molecular mechanism of the cell-cycle machinery in uterine leiomyoma has not yet been fully elucidated. Among the various types of cell-cycle regulators, p27(Kip1) (p27) is considered to be a potent tumor suppressor. To provide further molecular basis for understanding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service