Skip to Content
Merck
All Photos(1)

Key Documents

EHU010531

Sigma-Aldrich

MISSION® esiRNA

targeting human MMP9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTGACAGCGACAAGAAGTGGGGCTTCTGCCCGGACCAAGGATACAGTTTGTTCCTCGTGGCGGCGCATGAGTTCGGCCACGCGCTGGGCTTAGATCATTCCTCAGTGCCGGAGGCGCTCATGTACCCTATGTACCGCTTCACTGAGGGGCCCCCCTTGCATAAGGACGACGTGAATGGCATCCGGCACCTCTATGGTCCTCGCCCTGAACCTGAGCCACGGCCTCCAACCACCACCACACCGCAGCCCACGGCTCCCCCGACGGTCTGCCCCACCGGACCCCCCACTGTCCACCCCTCAGAGCGCCCCACAGCTGGCCCCACAGGTCCCCCCTCAGCTGGCCCCACAGGTCCCCCCACTGCTGGCCCTTCTACGGCCACTACTGTGCCTTTGAGTCCGGTGGACGATGCCTGCAACGTGAACATCTTCGACG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haibin Lu et al.
American journal of translational research, 9(12), 5361-5374 (2018-01-10)
Tumor-associated neutrophils (TANs) promote metastasis of multiple cancers, including oral squamous cell carcinoma (OSCC). Melatonin (Mel) reportedly exerts anti-metastatic effects on OSCC. However, little is known about the anti-OSCC effects of Mel involved in TANs. In this study, intensive infiltration
Hang Lin et al.
FEBS open bio, 7(9), 1291-1301 (2017-09-15)
Dysregulation of sirtuin 6 (SIRT6) is actively involved in tumor progression. High levels of SIRT6 have been associated with hepatocellular carcinoma and non-small cell lung cancer, and SIRT6 facilitates growth and metastasis of cancer cells. However, the clinical significance and
Xue Bai et al.
OncoTargets and therapy, 10, 2837-2847 (2017-06-28)
Epithelial-mesenchymal transition (EMT) is thought to be a crucial event during the early metastasis of tumor cells. Transforming growth factor (TGF)-β1 is involved in the process of EMT in a variety of human malignancies. Matrix metalloproteinase (MMP)-9 plays an important
Jon M Florence et al.
PloS one, 12(2), e0171427-e0171427 (2017-02-07)
The atherosclerotic process begins when vascular endothelial cells undergo pro-inflammatory changes such as aberrant activation to dysfunctional phenotypes and apoptosis, leading to loss of vascular integrity. Our laboratory has demonstrated that exposure of mice to second hand smoke triggers an
Jiaoli Wang et al.
Oncotarget, 8(47), 81880-81891 (2017-11-16)
Obesity is involved in tumor progression. However, the corresponding mechanisms remain largely unknown. Here, we report that adipocytes increase the invasive ability of tumor cells by producing exosomes with a high level of MMP3. Compared with 3T3-L1 cells, 3T3-L1 adipocytes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service