Skip to Content
Merck
All Photos(3)

Key Documents

E9406

Sigma-Aldrich

Epirubicin hydrochloride

≥90% (HPLC)

Synonym(s):

4′-Epidoxorubicin hydrochloride, Epidoxorubicin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29NO11 · HCl
CAS Number:
Molecular Weight:
579.98
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

biological source

synthetic

Quality Level

Assay

≥90% (HPLC)

form

powder

color

red to deep red

solubility

H2O: soluble

antibiotic activity spectrum

neoplastics

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

−20°C

SMILES string

Cl.COc1cccc2C(=O)c3c(O)c4C[C@](O)(C[C@H](O[C@H]5C[C@H](N)[C@@H](O)[C@H](C)O5)c4c(O)c3C(=O)c12)C(=O)CO

InChI

1S/C27H29NO11.ClH/c1-10-22(31)13(28)6-17(38-10)39-15-8-27(36,16(30)9-29)7-12-19(15)26(35)21-20(24(12)33)23(32)11-4-3-5-14(37-2)18(11)25(21)34;/h3-5,10,13,15,17,22,29,31,33,35-36H,6-9,28H2,1-2H3;1H/t10-,13-,15-,17-,22-,27-;/m0./s1

InChI key

MWWSFMDVAYGXBV-FGBSZODSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Epirubicin is used to inhibit topoisomerase II and DNA helicase activity. Epirubicin is used to study metastatic breast cancerand cardiac toxicity.

Biochem/physiol Actions

Epirubicin is antimitotic and cytotoxic. It inhibits nucleic acid and protein synthesis. Epirubicin may do so by forming complexes with DNA and intercalation between base pairs, by inhibiting topoisomerase II activity by stabilizing the DNA-topoisomerase II complex, and by preventing the religation portion of the ligation-religation reaction that topoisomerase II catalyzes.
Cell-permeable anthracycline antitumor antibiotic. Antineoplastic. A stereoisomer of doxorubicin that exhibits reduced cardiotoxicity. Its antitumor actions are mediated by targeting topoisomerase II.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Muta. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Letícia Tiburcio Ferreira et al.
Antimicrobial agents and chemotherapy, 64(9) (2020-07-01)
Widespread resistance against antimalarial drugs thwarts current efforts for controlling the disease and urges the discovery of new effective treatments. Drug repositioning is increasingly becoming an attractive strategy since it can reduce costs, risks, and time-to-market. Herein, we have used
O Feher et al.
Annals of oncology : official journal of the European Society for Medical Oncology, 16(6), 899-908 (2005-04-12)
This randomized, phase III study compared the efficacy and safety of first-line gemcitabine versus epirubicin in the treatment of postmenopausal women with metastatic breast cancer (MBC). Patients aged > or = 60 years (median 68 years) with clinically measurable MBC
D Groheux et al.
British journal of cancer, 109(5), 1157-1164 (2013-08-15)
Pathologic complete response (pCR) to neoadjuvant treatment (NAT) is associated with improved survival of patients with HER2+ breast cancer. We investigated the ability of interim positron emission tomography (PET) regarding early prediction of pathology outcomes. During 61 months, consecutive patients
Sarah N Hamilton et al.
International journal of radiation oncology, biology, physics, 87(4), 719-725 (2013-09-05)
To review the treatment and outcomes of patients with primary cutaneous B-cell lymphoma (CBCL). Clinical characteristics, treatment, and outcomes were analyzed for all patients referred to our institution from 1981 through 2011 with primary CBCL without extracutaneous or distant nodal
Yina Liao et al.
Cell death and differentiation, 28(4), 1347-1363 (2020-11-10)
CRSP8 plays an important role in recruiting mediators to genes through direct interaction with various DNA-bound transactivators. In this study, we uncovered the unique function of CRSP8 in suppressing thyroid cancer differentiation and promoting thyroid cancer progression via targeting IKKα

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service