Skip to Content
Merck
All Photos(1)

Key Documents

D9567

Sigma-Aldrich

Anti-Dopamine Transporter, C-Terminus antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DAT

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

monkey, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 1:1,000 using SDS-solubilized human striatal samples

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC6A3(6531)

Immunogen

synthetic peptide from the intracellular C-terminal region of human dopamine transporter.

Application

Anti-Dopamine Transporter, C-Terminus antibody produced in rabbit is suitable at a working dilution of 1:1000 for western blotting using SDS-solubilized human striatal samples and for immunohistochemistry using formaldehyde-fixed human and monkey brain sections.

Biochem/physiol Actions

The dopamine transporter (DAT1) acts to take released dopamine back up into presynaptic terminals and has been implicated in human disorders such as parkinsonism, Tourette syndrome, and substance abuse. DAT1 gene is a primary target gene for ADHD (Attention deficit hyperactivity disorder). DAT polymorphism along with maternal prenatal smoking is associated with childhood hyperactivity-impulsivity and inattentiveness.

Target description

The dopamine transporter (DAT1) acts to take released dopamine back up into presynaptic terminals and has been implicated in human disorders such as parkinsonism, Tourette syndrome, and substance abuse.

Physical form

Solution in 10 mM HEPES, pH 7.5, 150 mM NaCl, 100 μg/mL BSA, and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert S Kahn et al.
The Journal of pediatrics, 143(1), 104-110 (2003-08-14)
To examine the joint effects of a dopamine transporter (DAT) polymorphism and maternal prenatal smoking on childhood hyperactivity-impulsivity and inattentiveness. A cohort of 161 children was followed prospectively from age 6 months to 60 months. Primary outcomes were the DSM-IV
Theodora Papanikolaou et al.
The European journal of neuroscience, 29(12), 2264-2275 (2009-06-11)
The transcription factor Pitx3 is expressed exclusively by mesodiencephalic dopaminergic neurons; however, ablation of Pitx3 results in selective degeneration of primarily dopaminergic neurons of the substantia nigra pars compacta, the neuronal population that is most vulnerable in Parkinson's disease. Although
I D Waldman et al.
American journal of human genetics, 63(6), 1767-1776 (1998-12-05)
Attention-deficit hyperactivity disorder (ADHD) affects approximately 3%-5% of children in the United States. In the current psychiatric nomenclature, ADHD comprises three subtypes: inattentive, hyperactive-impulsive, and combined. In this study, we used four analytic strategies to examine the association and linkage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service