D7660
6-(γ,γ-Dimethylallylamino)purine
BioReagent, suitable for plant cell culture, 1 mg/mL
Synonym(s):
2iP, N6-(2-Isopentenyl)adenine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H13N5
CAS Number:
Molecular Weight:
203.24
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72
Recommended Products
sterility
sterile-filtered
Quality Level
product line
BioReagent
form
solution
concentration
1 mg/mL
technique(s)
cell culture | plant: suitable
application(s)
agriculture
shipped in
dry ice
storage temp.
−20°C
SMILES string
C\C(C)=C\CNc1ncnc2[nH]cnc12
InChI
1S/C10H13N5/c1-7(2)3-4-11-9-8-10(13-5-12-8)15-6-14-9/h3,5-6H,4H2,1-2H3,(H2,11,12,13,14,15)
InChI key
HYVABZIGRDEKCD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
6-(γ,γ-Dimethylallylamino)purine (2iP) is a bacteria-derived riboside cytokinin used to grow plant tissues such as tobacco and soybean callus. 2iP is a precursor of the cytokinin Zeatin. 2iP has been used in Schenk and Hildebrandt medium to support in vitro propagation of microshoot cultures from shoot tips of Genista plants.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J P Helgeson et al.
Plant physiology, 44(2), 193-198 (1969-02-01)
The growth rates of tobacco callus tissues on media containing 10(-6) to 10 mum 6-(gamma,gamma-dimethylallylamino)purine (2iP) were measured. At concentrations of 10(-4) mum and above growth rates were exponential and dependent on cytokinin concentration. At 2iP concentrations of 10(-4) to
G A Miura et al.
Plant physiology, 44(3), 372-376 (1969-03-01)
A 6-(gamma,gamma-dimethylallylamino) purine-like compound was found in the culture medium of Rhizopogon roseolus, which had been shown earlier to synthesize zeatin. The role of 6-(gamma,gamma-dimethylallylamino) purine as a precursor of zeatin was studied. Rhizopogon was furnished with 6-(gamma,gamma-dimethylallylamino) purine-8-(14)C. Cochromatography
E Strzelczyk et al.
Acta microbiologica Polonica, 34(2), 177-185 (1985-01-01)
Studies on the effect of post culture liquids of actinomycetes on cytokinin-like substances production by mycorrhizal fungi have revealed that actinomycete metabolites inhibited or stimulated the synthesis of these compounds. The results of chromatographic analyses suggest, that substances stimulating the
P W Hunt et al.
Plant molecular biology, 47(5), 677-692 (2001-12-01)
Haemoglobin genes have been found in a number of plant species, but the number of genes known has been too small to allow effective evolutionary inferences. We present nine new non-symbiotic haemoglobin sequences from a range of plants, including class
Yuki Ishii et al.
Biochimica et biophysica acta, 1745(2), 156-165 (2005-09-01)
Cytokinins are important purine derivatives that act as redifferentiation-inducing hormones to control many processes in plants. Cytokinins such as isopentenyladenine (IPA) and kinetin are very effective at inducing the granulocytic differentiation of human myeloid leukemia HL-60 cells. We examined the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service