Skip to Content
Merck
All Photos(1)

Key Documents

D7196

Sigma-Aldrich

2,5-Dimethyl-celecoxib

≥98% (HPLC)

Synonym(s):

2,5-Dimethylcelecoxib, 4-[5-(2,5-dimethylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]-benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16F3N3O2S
CAS Number:
Molecular Weight:
395.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

Cc1ccc(C)c(c1)-c2cc(nn2-c3ccc(cc3)S(N)(=O)=O)C(F)(F)F

InChI

1S/C18H16F3N3O2S/c1-11-3-4-12(2)15(9-11)16-10-17(18(19,20)21)23-24(16)13-5-7-14(8-6-13)27(22,25)26/h3-10H,1-2H3,(H2,22,25,26)

InChI key

NTFOSUUWGCDXEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,5-Dimethyl-celecoxib has been used
  • To block the synthesis of prostaglandin E2 and to study effect of transforming growth factor-2 signaling towards cyclooxygenase-2 expression.
  • In an in vitro study to analyze its pro-oxidative capacities in metastatic melanoma/ breast cancer cells.
  • To study its interaction between silymarin on chemically induced osteoarthritis in rats.

Biochem/physiol Actions

Celecoxib analog devoid of COX-2 inhibitor activity, induces apoptosis by inhibiting expression of survivin.
Celecoxib is a pyrazole derivative containing sulfonamide substituent. It is mostly metabolized by the cytochrome P450 2C9 system. Celecoxib possess anti-inflammatory action and can enhance antidepressant response.
Celecoxib, and the inactive analog 2′,5′-dimethyl celecoxib both induce apoptosis in a many cancer cell lines by down-regulating the expression of survivin. This activiy is mediated via an AKT mediated pathway, and is unique to this class of cyclo-oxygenase inhibitors, as non-selective NSAIDs or other COX-2 specific inhibitors such as rofecoxib, do not effect survivin expression. Unlike celecoxib, 2′,5′-dimethyl celecoxib is devoid of any inhibitory activity against COX-2.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trial of celecoxib in amyotrophic lateral sclerosis
Cudkowicz ME, et al.
Annals of Neurology, 60(1), 22-31 (2006)
Celecoxib inhibits mitochondrial O 2 consumption, promoting ROS dependent death of murine and human metastatic cancer cells via the apoptotic signalling pathway
Pritchard R, et al.
Biochemical Pharmacology (2018)
Clinical trial of adjunctive celecoxib treatment in patients with major depression: a double-blind and placebo controlled trial
Akhondzadeh S, et al.
Depression and Anxiety, 26(7), 607-611 (2009)
Oncology Nursing Drug Handbook, 521-521 (2004)
Meghan A Bowler et al.
JACC. Basic to translational science, 4(2), 135-143 (2019-05-08)
Calcific aortic valve disease is a progressive fibrocalcific process that can only be treated with valve replacement. Cadherin-11 has recently been identified as a potential therapeutic target for calcific aortic valve disease. The already approved drug celecoxib, a cyclooxygenase-2 inhibitor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service