Skip to Content
Merck
All Photos(1)

Key Documents

D6182

Sigma-Aldrich

DOTAP chloride

≥98% (TLC), powder

Synonym(s):

N-[1-(2,3-Dioleoyloxy)propyl]-N,N,N-trimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H80ClNO4
CAS Number:
Molecular Weight:
698.54
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98% (TLC)

form

powder

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(C[N+](C)(C)C)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Cl-]

InChI

1S/C42H80NO4.ClH/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-41(44)46-39-40(38-43(3,4)5)47-42(45)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2;/h20-23,40H,6-19,24-39H2,1-5H3;1H/q+1;/p-1/b22-20-,23-21-;

InChI key

KSXTUUUQYQYKCR-LQDDAWAPSA-M

Application

1,2-dioleoyloxy-3-(trimethylammonium)propane (DOTAP) chloride has been used for the preparation of cationic liposome and liposome-DNA complex. It has also been used to assess tumor necrosis factor α (TNF- α) production.
Cationic liposome-forming compound for transfection of DNA, RNA and other negatively charged molecules into eukaryotic cells. Mixing DOTAP with DNA results in spontaneously formed stable complexes that can be directly added to cell culture medium. These complexes fuse with the cell membrane and release DNA into the cytoplasm. The cells are transfected efficiently without cytotoxic effects. Use approximately 5-10 μg DOTAP per μg DNA and approximately 10-20 μg DOTAP per mL cell culture medium. The optimal working concentration and transfection time depends in part on the cell line used, and the type of material (RNA, DNA, protein) loaded.

Biochem/physiol Actions

1,2-dioleoyloxy-3-(trimethylammonium)propane (DOTAP) chloride is useful in liposome preparation, without the use of helper lipid. DOTAP chloride is soluble in dimethyl sulfoxide (DMSO) and ethanol, this defines its function as a transfection agent.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structurally distinct phosphatases CD45 and CD148 both regulate B cell and macrophage immunoreceptor signaling
Zhu JW, et al.
Immunity, 28(2), 183-196 (2008)
Ju-mei Zhao et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 22(12), 4272-4280 (2008-08-22)
The matrix (M) protein of vesicular stomatitis virus (VSV) plays a key role in inducing cell apoptosis during infection. To investigate whether M protein-mediated apoptosis could be used in cancer therapy, its cDNA was amplified and cloned into eukaryotic expression
A novel strategy for tumour therapy combining cell apoptosis and active immunity induced by caspy2, a zebrafish caspase
Liu L, et al.
Journal of Cellular and Molecular Medicine, 13(8b), 2271-2281 (2009)
A simplified direct lipid mixing lipoplex preparation: Comparison of liposomal-, dimethylsulfoxide-, and ethanol-based methods
Meisel JW and Gokel GW
Scientific Reports, 6, 27662-27662 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service