Skip to Content
Merck
All Photos(1)

Key Documents

D5919

Sigma-Aldrich

Diacylglycerol Kinase Inhibitor I

solid

Synonym(s):

6-[2-[4-[(4-Fluorophenyl)phenylmethylene]-1-piperidin­yl]ethyl]-7-methyl-5H-thiazolo-[3,2-a]-pyrimidin-5-one, R59022

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H26FN3OS
CAS Number:
Molecular Weight:
459.58
MDL number:
UNSPSC Code:
41106300
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥98% (HPLC)

form

solid

color

pale yellow

solubility

0.1 M HCl: slightly soluble
0.1 M NaOH: slightly soluble
DMSO: soluble
H2O: insoluble
ethanol: soluble
ethyl acetate: soluble

storage temp.

−20°C

SMILES string

CC1=C(CCN2CCC(\CC2)=C(\c3ccccc3)c4ccc(F)cc4)C(=O)N5C=CSC5=N1

InChI

1S/C27H26FN3OS/c1-19-24(26(32)31-17-18-33-27(31)29-19)13-16-30-14-11-22(12-15-30)25(20-5-3-2-4-6-20)21-7-9-23(28)10-8-21/h2-10,17-18H,11-16H2,1H3

InChI key

MFVJXLPANKSLLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diacylglycerol Kinase Inhibitor I prevents the formation of foreign body giant cell (FBGC). It inhibits the phosphorylation of diacylglycerol to phosphatidic acid. Diacylglycerol Kinase Inhibitor I functions as a serotonin (5-HT) receptor (5-HTR) antagonist. It stimulates apoptosis and autophagy in neuronal cell line NG108-15.

Application

Diacylglycerol Kinase Inhibitor I has been used as an inhibitor to study suppression of T cell function using advanced generation human CAR (chimeric antigen receptors) T cells.

Biochem/physiol Actions

Diacylglycerol kinase inhibitor. Inhibits formation of [38P]1-Oleoxyl-2-acetylglyceryl-3-phosphoric acid (OAPA) in red blood cell membranes: IC50 = 3.3 μM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activation of phospholipase C and protein kinase C has little involvement in ADP-induced primary aggregation of human platelets: effects of diacylglycerols, the diacylglycerols, the diacylglycerol kinase inhibitor R59022, staurosporine and okadaic acid
Packham MA, et al.
The Biochemical Journal, 290(3), 849-856 (1993)
Eric Ruelland et al.
Plant physiology, 130(2), 999-1007 (2002-10-12)
The signaling events generated by a cold exposure are poorly known in plants. We were interested in checking the possible activation of enzymes of the phosphoinositide signaling pathway in response to a temperature drop. In Arabidopsis suspension cells labeled with
Multifactorial T-cell hypofunction that is reversible can limit the efficacy of chimeric antigen receptor-transduced human T cells in solid tumors
Moon EK, et al.
Clinical Cancer Research, 197-201 (2014)
Amy K McNally et al.
The American journal of pathology, 163(3), 1147-1156 (2003-08-26)
Multinucleated foreign body giant cells (FBGCs) form by monocyte-derived macrophage fusion on implanted biomedical devices and are believed to mediate oxidative damage to biomaterial surfaces. Our in vitro system of human macrophage culture and interleukin (IL)-4-induced FBGC formation was developed
Ana M Laxalt et al.
The Journal of biological chemistry, 282(29), 21160-21168 (2007-05-11)
Nitric Oxide (NO) is a second messenger related to development and (a)biotic stress responses in plants. We have studied the role of NO in signaling during plant defense responses upon xylanase elicitation. Treatment of tomato cell cultures with the fungal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service