Skip to Content
Merck
All Photos(2)

Key Documents

D5679

Sigma-Aldrich

Methyl all-cis-7,10,13,16,19-docosapentaenoate

≥97%, liquid

Synonym(s):

cis-7,10,13,16,19-Docosapentaenoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H36O2
CAS Number:
Molecular Weight:
344.53
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥97%

form

liquid

functional group

ester

lipid type

omega FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCCCC(=O)OC

InChI

1S/C23H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23(24)25-2/h4-5,7-8,10-11,13-14,16-17H,3,6,9,12,15,18-22H2,1-2H3/b5-4-,8-7-,11-10-,14-13-,17-16-

InChI key

PTFHIRHGARALFY-JEBPEJKESA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kun Kang et al.
Veterinary medicine and science, 6(4), 755-765 (2020-06-27)
This research aimed to investigate the effects of dietary energy concentration (combined net energy, Nemf) on growth performance and meat quality of yaks raised by barn feeding. In all, 30 male yaks (3-year old and 114.57 ± 21.56 kg of body weight) were
Edgar Weigand et al.
Lipids, 47(3), 291-302 (2011-12-01)
The impact of a moderate Zn deficiency on growth and plasma and liver lipids was investigated in two 4-week experiments with male weanling rats fed fat-enriched diets. Semisynthetic, approximately isocaloric diets containing 3% soybean oil were supplemented with either 7
Robert Bodkowski et al.
Animals : an open access journal from MDPI, 10(10) (2020-10-15)
The study aimed to investigate the effect of supplementation of fish oil after the process of low-temperature crystallization (LTC-FO) enriched with long-chain polyunsaturated fatty acids (LC-PUFAs) on cow milk parameters. The experiment was carried out on 24 Polish Holstein Friesian

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service