Skip to Content
Merck
All Photos(1)

Key Documents

D3822

Sigma-Aldrich

Dipeptidyl Peptidase IV Inhibitor I

Synonym(s):

Diprotin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H31N3O4
Molecular Weight:
341.45
UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

solid

color

white

storage temp.

2-8°C

General description

Dipeptidyl peptidase IV (DPP IV) is a member of the prolyl-specific proteases family. It is a dimeric type II integral membrane glycoprotein that is broadly expressed in the epithelial and nonepithelial tissues. It is majorly expressed in the kidney and the colon. 

Application

Dipeptidyl peptidase IV inhibitor I has been used in acute in vivo studies to assess rapid plasma glucagon-like peptide (GLP-1) responses to specified secretory agents in mice.

Biochem/physiol Actions

Dipeptidyl peptidase IV (DPP IV) functions as a modulator of glucagon-like peptide (GLP-1), insulin-stimulating hormones, and glucose-dependent insulinotropic polypeptide (GIP). It is an essential therapeutic target for type 2 diabetes (T2DM). DPP-4 inhibitors are crucial for maintaining glucose homeostasis by prolonging the activity of GLP-1 and GIP. They also enhance glycemic control for a longer period when compared to early oral hypoglycemics. DPP-4 also acts as a binding protein and a ligand of extracellular factors.

Sequence

Ile-Pro-Ile

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco Alejandro Lagunas-Rangel et al.
Biomedicines, 11(11) (2023-11-25)
Dipeptidyl peptidase 4 (DPP4) inhibitors, commonly known as gliptins, have been an integral part of the treatment of type 2 diabetes mellitus (T2DM) for several years. Despite their remarkable efficacy in lowering glucose levels and their compatibility with other hypoglycemic
Sebastiano Bruno Solerte et al.
Acta diabetologica, 57(7), 779-783 (2020-06-09)
SARS-CoV-2 causes severe respiratory syndrome (COVID-19) with high mortality due to a direct cytotoxic viral effect and a severe systemic inflammation. We are herein discussing a possible novel therapeutic tool for COVID-19. Virus binds to the cell surface receptor ACE2;
Rajesh Gupta et al.
Current drug targets, 10(1), 71-87 (2009-01-20)
Dipeptidyl peptidase IV (DPP IV) is a key regulator of insulin-stimulating hormones, glucagon-like peptide (GLP-1) and glucose-dependent insulinotropic polypeptide (GIP), thus it is a promising target for the treatment of Type 2 Diabetes mellitus (T2DM). Inhibition of plasma DPP IV
Brandon L Panaro et al.
Molecular metabolism, 37, 100990-100990 (2020-04-13)
Glucagon-like peptide-1 is a nutrient-sensitive hormone secreted from enteroendocrine L cells within the small and large bowel. Although GLP-1 levels rise rapidly in response to food ingestion, the greatest density of L cells is localized to the distal small bowel
Sourav De et al.
Mini reviews in medicinal chemistry, 19(2), 88-97 (2018-04-26)
Diabetes mellitus is an emerging predator and affecting around 422 million adults worldwide. Higher levels of circulating insulin and increased pressure on the pancreas to produce insulin have been inferred as possible etiology for diabetes leading to a higher risk

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service