Skip to Content
Merck
All Photos(1)

Key Documents

D3658

Sigma-Aldrich

1,9-Dideoxyforskolin from Coleus forskohlii

≥97%, solid

Synonym(s):

7β-Acetoxy-6β-hydroxy-8,13-epoxy-labd-14-en-11-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O5
CAS Number:
Molecular Weight:
378.50
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97%

form

solid

optical activity

[α]26/D +93.6°, c = 6.12 in chloroform(lit.)

color

white to off-white

solubility

methanol: 28 mg/mL(lit.)
DMSO: 3 mg/mL(lit.)
chloroform: 50 mg/mL
ethanol: 6.6 mg/mL(lit.)
dilute aqueous acid and base: insoluble(lit.)

storage temp.

−20°C

SMILES string

[H][C@@]12[C@H](O)[C@H](OC(C)=O)[C@@]3(C)O[C@](C)(CC(=O)C3[C@@]1(C)CCCC2(C)C)C=C

InChI

1S/C22H34O5/c1-8-20(5)12-14(24)16-21(6)11-9-10-19(3,4)17(21)15(25)18(26-13(2)23)22(16,7)27-20/h8,15-18,25H,1,9-12H2,2-7H3/t15-,16?,17-,18-,20-,21+,22-/m0/s1

InChI key

ZKZMDXUDDJYAIB-OJPJTMFRSA-N

Application

Useful as a negative control for forskolin.

Biochem/physiol Actions

Biologically inactive forskolin analog. Does not stimulate adenylyl cyclase.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Adenylyl cyclases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P d'Alcantara et al.
Biophysical journal, 77(1), 204-216 (1999-07-02)
The activity of the voltage-gated Na+ channel is subjected to modulation through covalent modifications. It has been previously shown that brain Na+ currents are reduced following the activation of the protein kinase A (PKA) pathway, but the effect of the
C Y Liu et al.
Brain research, 826(2), 253-269 (1999-05-04)
The aims of this study were to improve insight into cAMP signaling in myenteric neurons and glia and identify the adenylyl cyclase (AC) isoforms expressed in myenteric ganglia of the guinea-pig small intestine. An increase in the intracellular cAMP levels
Hisahiro Hagiwara et al.
The Journal of organic chemistry, 71(12), 4619-4624 (2006-06-06)
Forskolin (1), a highly oxygenated labdane diterpenoid and an activator of adenylate cyclase, has been synthesized in 12 steps and 12% overall yield from ptychantin A (4), which has been isolated from liverwort Ptychanthus striatus in good yield. The 1alpha-hydroxy
Y P Vedernikov et al.
American journal of obstetrics and gynecology, 182(3), 620-624 (2000-03-30)
We sought to study the contribution of potassium channels in the effect of forskolin and 1,9-dideoxyforskolin on uterine contractility in the pregnant rat. Rings taken from the middle portions of uterine horns from rats at 16 days of gestation were
K Schmidt et al.
Journal of cardiovascular pharmacology, 13(3), 353-360 (1989-03-01)
The effects of forskolin and seven derivatives on cardiac functions were investigated by using the Langendorff technique and the results compared with the respective potencies obtained from adenylate cyclase and binding studies. In the isolated heart, forskolin increased all parameters

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service