Skip to Content
Merck
All Photos(1)

Key Documents

D2916

Sigma-Aldrich

Lambda DNA Mixed Digest

for DNA electrophoresis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

100
200

form

buffered aqueous solution

usage

100 uses (0.1 mg size)
20 uses (20 μg size)

suitability

suitable for electrophoresis (DNA)

storage temp.

−20°C

General description

Sigma′s Lambda DNA Mixed Digest contains 10 fragments from 1,503 − 48,502 bp. Six ul of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel.

Application

Lambda DNA Mixed Digest has been used for DNA electrophoresis.
Suitable for size determination of dsDNA by electrophoresis with either agarose or polyacrylamide gels.

Components

Sigma′s Lambda DNA Mixed Digest is provided in a solution of 10 mM Tris-HCl (pH 8.0), with 1.0 mM EDTA.

Preparation Note

Bacteriophage lambda DNA was completely digested with by Apa I, Kpn I, Xba I and Xho I in separate reactions, sticky ends filled with DNA polymerase I Klenow (to prevent reannealing), mixed with uncut bacteriophage lambda DNA, and then purified by phenol extraction.

Other Notes

For optimal resolution, the recommended agarose gel concentration is 0.4%.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A rapid (100 min) method for isolating high yield and quality DNA from leaves, roots and coleoptile of wheat (Triticum aestivum L.) suitable for apoptotic and other molecular studie
Hameed
International Journal of Agriculture and Biology, 383-387 (2004)

Articles

Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.

Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.

Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.

Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service