Skip to Content
Merck
All Photos(1)

Key Documents

D218

Sigma-Aldrich

Monoclonal Anti-Dihydropyridine Receptor (α1 Subunit) antibody produced in mouse

clone 1A, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

conjugate

unconjugated

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1A, monoclonal

form

buffered aqueous solution

mol wt

antigen ~200 kDa

species reactivity

human (weakly), guinea pig, mouse, rat, rabbit

technique(s)

immunohistochemistry (frozen sections): 1:200
immunoprecipitation (IP): suitable
western blot (chemiluminescent): 1:500

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

DHP receptor is composed of at least four subunits. The α1 subunit contains the binding site for the DHPs and shows high sequence homology to the voltage gated Na+ channel. The α2 subunit is a large glycoprotein and the other 2 subunits are termed β and γ.

Specificity

Reacts specifically with the α1 subunit of the DHP receptor. May be used to inhibit the DHP-sensitive calcium current in BC3H1 mouse muscle cells. By immunohistochemistry, double rows of discrete punctate staining represent pairs of triads on the opposing sides of the Z-lines.

Immunogen

rabbit muscle transverse tubule dihydropyridine (DHP) receptors.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Monoclonal Anti-Dihydropyridine Receptor (α1 Subunit) antibody produced in mouse is suitable for immunoprecipitation, immunohistochemistry at a working dilution of 1:500 and immunoblotting at 1:2000. It was used for the detection and localization of DHPR-1α by western blotting in skeletal muscle tissue sections of mice in a study. The antibody may be used to inhibit the DHP-sensitive calcium current in BC3H1 mouse muscle cells.

Biochem/physiol Actions

The 1,4-dihydropyridines (DHPs) are synthetic organic compounds which can be used to identify the L-type Ca2+ channels that are found in all types of vertebrate muscle, neuronal and neuroendocrine cells. DHP receptor is part of the L-type calcium channel complex and is thought to be the voltage sensor in E-C coupling.

Physical form

Solution in phosphate buffered saline containing 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Van et al.
FEBS letters, 589(10), 1095-1104 (2015-04-07)
Mitsugumin 56 (MG56), also known as the membrane-bound O-acyl-transferase family member hedgehog acyltransferase-like, was identified as a new sarcoplasmic reticulum component in striated muscle. Mg56-knockout mice grew normally for a week after birth, but shortly thereafter exhibited a suckling defect
F Galbiati et al.
The Journal of biological chemistry, 276(24), 21425-21433 (2001-03-22)
Caveolin-3, a muscle-specific caveolin-related protein, is the principal structural protein of caveolae membrane domains in striated muscle cells. Recently, we identified a novel autosomal dominant form of limb-girdle muscular dystrophy (LGMD-1C) in humans that is due to mutations within the
Pankaj B Agrawal et al.
American journal of human genetics, 95(2), 218-226 (2014-08-05)
Centronuclear myopathies (CNMs) are characterized by muscle weakness and increased numbers of central nuclei within myofibers. X-linked myotubular myopathy, the most common severe form of CNM, is caused by mutations in MTM1, encoding myotubularin (MTM1), a lipid phosphatase. To increase
Philippe Magown et al.
Nature communications, 6, 8506-8506 (2015-10-16)
Neural prostheses can restore meaningful function to paralysed muscles by electrically stimulating innervating motor axons, but fail when muscles are completely denervated, as seen in amyotrophic lateral sclerosis, or after a peripheral nerve or spinal cord injury. Here we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service