Skip to Content
Merck
All Photos(1)

Key Documents

D204

Sigma-Aldrich

S-(+)-O-Desmethylraclopride hydrobromide

solid, ≥98% (HPLC)

Synonym(s):

(S)-3,5-Dichloro-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2,6-dihydroxybenzamide hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18Cl2N2O3 · HBr
CAS Number:
Molecular Weight:
414.12
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

optical activity

[α]23/D +11.0°, c = 1.08 in ethanol(lit.)

storage condition

protect from light

color

white

solubility

H2O: 10 mg/mL

SMILES string

Br.CCN1CCC[C@H]1CNC(=O)c2c(O)c(Cl)cc(Cl)c2O

InChI

1S/C14H18Cl2N2O3.BrH/c1-2-18-5-3-4-8(18)7-17-14(21)11-12(19)9(15)6-10(16)13(11)20;/h6,8,19-20H,2-5,7H2,1H3,(H,17,21);1H/t8-;/m0./s1

InChI key

RIWAHQQBHAKCTN-QRPNPIFTSA-N

Application

Precursor to radiolabeled S-(+)-raclopride.

Caution

Light sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Adam et al.
International journal of radiation applications and instrumentation. Part A, Applied radiation and isotopes, 40(1), 91-92 (1989-01-01)
This paper describes the separation of lipophilic amine radiopharmaceuticals on normal phase silica using a reversed phase type eluant. This class of compound usually gives very poor peak symmetry on reversed phase columns. However, excellent separation of a number of
V W Pike et al.
International journal of radiation applications and instrumentation. Part A, Applied radiation and isotopes, 41(5), 483-492 (1990-01-01)
Practical methods are described for the quality assurance of three labelled agents (L-6-[18F]fluoro-DOPA, S-[N-methyl-11C]nomifensine and [O-methyl-11C]raclopride) now produced regularly for PET studies of the dopaminergic system in man. These include indirect methods for the initial determination of label position (e.g.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service