CMC0007
SIG10 5a Chemically Competent Cells
Escherichia coli, rod shaped
Synonym(s):
Chemically Competent Cells
About This Item
Recommended Products
product name
SIG10 5a Chemically Competent Cells, for DNA plasmid production
biological source
Escherichia coli
grade
for molecular biology
form
buffered aqueous solution
growth mode
adherent or suspension
morphology
rod shaped
technique(s)
microbiological culture: suitable
cell transformation
competent cell type: chemically competent
transformation efficiency: ≥1 x 108 cfu/μg
shipped in
dry ice
storage temp.
−70°C
1 of 4
This Item | EP4925000162 | EP4925000022 | EP4924000010 |
---|---|---|---|
single-channel, fixed | single-channel, fixed | single-channel, fixed | includes epT.I.P.S.® box, single-channel, variable |
(2.5 mL) | (2 mL) | (2 μL) | (0.1-2.5 μL) |
pack of 1 ea | pack of 1 ea | pack of 1 ea | pack of 1 ea |
red | red | dark gray | dark gray |
General description
They provide the performance researchers need with ease of use as only common laboratory equipment is required and provide solutions for a wide range of applications at economical prices.
Genotype
fhuA2Δ(argF-lacZ)U169 phoA glnV44 Φ80 Δ(lacZ)M15 gyrA96 recA1 relA1 endA1 thi-1 hsdR17
Features and Benefits
- Direct replacements for standard cloning strains like DH5α™, DH10B, JM109, TOP10, XL1-Blue, etc.
- Optimized genetics for high yields: endonuclease and recombination minus
- Blue - white screening
Components
- SIG10 5a chemically competent cells
- pUC 19 transformation control DNA
- recovery medium for cloning
Legal Information
related product
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service