Key Documents
CLS6749
Corning® Cell Counter
Synonym(s):
Automated counter
About This Item
UNSPSC Code:
42181504
NACRES:
NC.01
material
aluminum
description
USB 3.0
packaging
pkg of 1 ea
manufacturer/tradename
Corning 6749
H × L × W
(100 mm) (90 mm) (90 mm)
weight
1.0 kg
Related Categories
General description
For years, the choice between manual and automated cell counting has been a difficult one. Manual cell counting, on the one hand, is accurate, but time-consuming and very user-dependent. Automated cell counting, is much faster and less user-dependent, but the cost of disposable counting slides can be an issue. A tough choice, but now there is a solution.
The new Corning Cell Counter is the first automated cell counter that combines the best of both worlds. It is:
  • Fast – thanks to its online image processing
  • Accurate – thanks to its cloud-based machine learning algorithm.
  • Low-cost – works with common reusable glass hemocytometer. No consumables required.
System Requirements
Windows 10 tablet or computer with a USB 3.0 port.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1
1 of 1
Related Content
This guide helps determine if an open or closed system is the best method for your Corning® cell counting strategy.
This guide covers how Corning® Automated Cell Counters are able to count cells using the cloud