CLS6569
Corning® microplate sealing tape
aluminum film, non-sterile, pack of 100
Synonym(s):
microplate sealing film, multiwell plate sealing film, sealing film
About This Item
Recommended Products
material
aluminum film
sterility
non-sterile
feature
piercable
packaging
pack of 100
manufacturer/tradename
Corning 6569
size
384 wells
color
none
suitability
suitable for (for short and long term storage and PCR, 384 well reactions)
Looking for similar products? Visit Product Comparison Guide
General description
- Easy application and removal for short- and long-term storage
- Provide tight seal to minimize evaporation and condensation and enables oil-free operation when used with PCR thermal cyclers with heated lids
- Suitable for use between -80°C and 150°C, is not transparent, and is pierceable
- DMSO-resistant
- Nonsterile
- Certified DNase- and RNase-free
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service