CLS5641P250
Corning® reusable volumetric flask, Class B
polypropylene, size 250 mL, with 19/26 tapered PP stopper
Synonym(s):
Corning container
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
long
polypropylene
sterility
non-sterile
feature
class B
closure type tapered polypropylene stopper
packaging
pack of 1
case of 5
manufacturer/tradename
Corning 5641P-250
H × O.D.
225 mm × 84 mm
size
250 mL
Looking for similar products? Visit Product Comparison Guide
General description
Transluscent flask is supplied with tapered polypropylene stopper with an individually calibrated graduation line. The product is manufactured to Class B tolerances as defined by ISO 1042. Exposure to temperatures above 60 °C may affect accuracy. Recommended routine cleaning up to 60 ºC will preserve graduations. Height dimension is for flask only, without stopper.
Legal Information
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service