Key Documents
CLS430293
Corning® tissue-culture treated culture dishes
D × H 100 mm × 20 mm
Synonym(s):
petri dish
About This Item
UNSPSC Code:
41121812
NACRES:
NB.13
Pricing and availability is not currently available.
material
polystyrene
round
sterility
sterile
usage
11.0-16.5 mL Optimal culture medium volume range (Optimal culture medium volume)
packaging
sleeve of 10
manufacturer/tradename
Corning 430293
D × H
100 mm × 20 mm
surface area
55 cm2
Features and Benefits
  • Manufactured from optically clear virgin polystyrene
  • Treated for optimal cell attachment
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Lid is supplied with vents to provide consistent gas exchange
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)