Skip to Content
Merck
All Photos(1)

Key Documents

CLS4130

Corning® DeckWorks pipet tips reload starter kits

0.1 - 10μL pipet tips, graduated, natural, non-sterile, polypropylene, 96 tips/deck, 10 decks/pack, 960 tips/pack, 1 pack/case and 10 empty racks/case

Synonym(s):

pipette tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume range:
0.1-10 μL
sterility:
non-sterile
feature:
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Tip format: 96 tips/rack
Tip shape: round
packaging:
rack of 96
manufacturer/tradename:
Corning 4130
Pricing and availability is not currently available.

material

natural tip
polypropylene

description

Starter Kit

sterility

non-sterile

feature

DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Tip format: 96 tips/rack
Tip shape: round

packaging

rack of 96

manufacturer/tradename

Corning 4130

volume range

0.1-10 μL

Looking for similar products? Visit Product Comparison Guide

General description

Corning® DeckWorks Reload Starter Kits

DeckWorks starter kits contain one DeckWorks reload system with standard tips and empty hinged racks. 10 empty racks accompany 10 uL, 200 uL and 300 uL volume tips and 8 empty racks accompany 1000 uL volume tips.

This system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials. Both the hinged racks and reload decks within this system are manufactured with recycled plastic.

The pipet tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes.

Medical grade resins and production excellence allow for full certification as:

  • Nonpyrogenic
  • Human DNA-free
  • RNase-/DNase-free

Legal Information

Corning is a registered trademark of Corning, Inc.
DeckWorks is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service