Key Documents
CLS402614
Corning® direct overhead drive assembly for ProCulture® spinner flasks
direct overhead drive, 100 mm center cap assembly, 1/cs
Synonym(s):
Corning container
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
sterility
non-sterile
packaging
pack of 1
case of 2
manufacturer/tradename
Corning 402614
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
CLS45158LCLS451515LCLS451536L
packaging
pack of 1
packaging
case of 1, pack of 1
packaging
case of 1, pack of 1
packaging
case of 1, pack of 1
manufacturer/tradename
Corning 402614
manufacturer/tradename
Corning 4515-8L
manufacturer/tradename
Corning 4515-15L
manufacturer/tradename
Corning 4515-36L
General description
100mm center cap assembly for Corning® Proculture® spinner flasks is designed for use with direct overhead drive motor. These cap assemblies required optional 8L, 15L or 36L paddle assemblies and can be used on all 4500 and 4510 series vertical sidearm flasks.
Legal Information
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries