Skip to Content
Merck
All Photos(1)

Key Documents

CLS3903

Corning® 96 Well White Polystyrene Microplate

flat bottom clear, white polystyrene, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear bottom
flat bottom clear
white polystyrene
size:
96 wells
surface area:
0.32 cm2 , cell growth area

cell growth area

cell growth area
well volume:
360 μL
working volume:
75-200 μL

material

clear bottom
flat bottom clear
white polystyrene

sterility

sterile

feature

lid
skirt

packaging

case of 100
bag of 20

manufacturer/tradename

Corning 3903

size

96 wells

surface area

0.32 cm2 , cell growth area

cell growth area

cell growth area

well volume

360 μL

working volume

75-200 μL

color

white

suitability

suitable for (cell culture)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning® 96 Well White Polystyrene Microplate are sterile polystyrene plates with clear, flat, white bottom. It is equipped with non-reversible lids featuring condensation rings to ensure a secure seal. Moreover, the alphanumeric codes help in easy well identification. These microplates are treated to provide optimal cell attachment, promoting successful cell culture experiments.

Application

Corning® 96 Well White Polystyrene Microplate has been used in single-cycle infectivity assays.

Features and Benefits

  • Flat, optically clear well bottom with white painted wall plates enables direct microscopic examination
  • The presence of condensation rings on non-reversible lids minimizes contamination
  • Gamma radiation sterilized and nonpyrogenic in nature
  • Presence of opaque walls to avoid crosstalk between wells
  • White-colored walled polystyrene microplates boost luminescent signals while reducing fluorescence and background luminescence
  • 60% thinner bottoms which allow reading as low as 340nm
  • Can be used for well identification due to the presence of Individual alphanumeric codes
  • Used as direct microscopic observation due to the presence of a clear well with a flat bottom
  • Preferred for optimal cell attachment
  • Suitable for the bottom and top reading instruments

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elucidating the basis for permissivity of the MT-4 T-cell line to replication of an HIV-1 mutant lacking the gp41 cytoplasmic tail
Melissa V Fernandez
Journal of Virology, 94, e01334-e01320 (2020)
Xingrui He et al.
Royal Society open science, 7(5), 200038-200038 (2020-06-17)
Herein, we report the first access of β-elemene derivatives through the SeO2-mediated oxidation reaction. Several new compounds were isolated through such a one-step reaction, and their structures were elucidated using various 2D-NMR techniques. This method provides easy access to multiple

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service