Key Documents
CLS3883
Corning® 96 Well Half-Area Microplate
flat bottom clear, white polystyrene, bag of 25, non-sterile, lid: no
Synonym(s):
96 multiwell plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
Pricing and availability is not currently available.
material
flat bottom clear
white polystyrene
description
non-treated
sterility
non-sterile
feature
lid: no
packaging
bag of 25
pkg of 25 inner package
manufacturer/tradename
Corning 3883
well D
10.7 mm
General description
Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.
Legal Information
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Need A Sample COA?
This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.