CLS3732
Corning® CellSTACK® accessories
universal cap, diam. 33 mm, sterile
Synonym(s):
cell culture chamber accessory
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
screw top orange high-density polyethylene cap
sterility
sterile
packaging
pack of 4 ea
manufacturer/tradename
Corning 3732
diam.
33 mm
Looking for similar products? Visit Product Comparison Guide
General description
- 33 mm polyethylene threaded cap
- Vented overcap has 0.2 μm PTFE membrane sealed into the peelable foil cover
- Sterilized by gamma irradiation
- Double bagged
Available for filling, venting, stacking and manipulation.
Legal Information
CellSTACK is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service