Skip to Content
Merck
All Photos(1)

Key Documents

CLS356701

Corning® BioCoat Collagen I Microplate

well size 96 wells, flat bottom clear white wells

Synonym(s):

Cell Culture Plate, plastic coated cell culture plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.13
material:
flat bottom clear white wells
size:
96 well
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid (Clear polystyrene lid)

material

flat bottom clear white wells

sterility

sterile

feature

lid (Clear polystyrene lid)

packaging

pkg of 80 ea (4 packs of 80)

manufacturer/tradename

Corning 356701

size

96 well

well size

96 wells

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Application

Cell culture

Legal Information

BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service