CLS353089
Corning® Falcon® Cell Scraper
blade L 3.0 cm, handle L 25 cm, sterile
Synonym(s):
lab equipment accessory
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
material
TPE
sterility
sterile
manufacturer/tradename
Corning 353089
technique(s)
cryopreservation: suitable
blade L
3.0 cm
handle L
25 cm
Looking for similar products? Visit Product Comparison Guide
Legal Information
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service