Key Documents
CLS3294
Corning® CellBIND® surface culture dishes
D × H 35 mm × 10 mm
Synonym(s):
plastic cell culture dish
About This Item
UNSPSC Code:
41121812
NACRES:
NB.13
Pricing and availability is not currently available.
material
polystyrene
round
sterility
sterile; γ-irradiated
packaging
sleeve of 10
manufacturer/tradename
Corning 3294
D × H
35 mm × 10 mm
surface area
8 cm2
General description
  • Manufactured from optically-clear virgin polystyrene
  • Corning® CellBIND® surface treated for optimal cell attachment
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Supplied with vents to provide consistent gas exchange
Legal Information
CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
For good cell attachment the hydrophobic polystyrene surface must be modified to a more hydrophilic surface. This allows cell attachment proteins (vitronectin and fibronectin) found in the serum containing culture medium to adhere and spread on the vessel bottom providing a better surface for cells to attach
For good cell attachment the hydrophobic polystyrene surface must be modified to a more hydrophilic surface. This allows cell attachment proteins (vitronectin and fibronectin) found in the serum containing culture medium to adhere and spread on the vessel bottom providing a better surface for cells to attach
For good cell attachment the hydrophobic polystyrene surface must be modified to a more hydrophilic surface. This allows cell attachment proteins (vitronectin and fibronectin) found in the serum containing culture medium to adhere and spread on the vessel bottom providing a better surface for cells to attach
For good cell attachment the hydrophobic polystyrene surface must be modified to a more hydrophilic surface. This allows cell attachment proteins (vitronectin and fibronectin) found in the serum containing culture medium to adhere and spread on the vessel bottom providing a better surface for cells to attach