CLS2019
Corning® cap inserts for cryogenic vials
yellow
Synonym(s):
Corning cap
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
sterility
non-sterile
manufacturer/tradename
Corning 2019
color
yellow
Looking for similar products? Visit Product Comparison Guide
General description
- Cap inserts provide color coding for easy sample identification
- Inserts are packaged in resealable bags
- Cap inserts fit all Corning cryogenic vials except Corning No. 430289
Legal Information
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service