CHA00011
EUR-2019-nCoV Assays for detection of SARS-CoV-2 RNA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105500
NACRES:
NA.51
Recommended Products
form
solution (in 10 mM Tris, 1 mM EDTA)
packaging
pkg of 2 vials
pH range
7.5-8.5
shipped in
ambient
storage temp.
−20°C
General description
EUR-2019-nCoV Assays are for qPCR (5′ nuclease assay) detection of SARS-CoV-2.
The product is primers and probes, the sequences of which were designed and made publicly available by Charité Institute of Virology – Universitätsmedizin Berlin.
For additional details, please visit SigmaAldrich.com/ncov.
The product is primers and probes, the sequences of which were designed and made publicly available by Charité Institute of Virology – Universitätsmedizin Berlin.
For additional details, please visit SigmaAldrich.com/ncov.
Features and Benefits
The forward and reverse primers in each tube come with the following features:
The Dual-Labeled Probe in each tube comes with the following features:
- DNA bases
- No modifications
- Desalt purified
- 8 nmol each E_Sarbeco & 10 nmol each Modified RdRp
The Dual-Labeled Probe in each tube comes with the following features:
- DNA bases
- 6-FAM™ reporter
- BXQ (Block All Quencher—SigmaAldrich.com/bxq)
- HPLC purified
- 4 nmol E_Sarbeco & 5 nmol Modified RdRp
Components
The product includes 2 individual tubes of 20X primer and probe pools for quantification of SARS-CoV-2 genes. Assays included are E_Sarbeco and Modified RdRp (formerly Discrimatory) genes.
The forward primer, reverse primer, and probe are at final concentrations of 8, 8, and 4 μM (E_Sarbeco); and, 10, 10, and 5 μM (Modified RdRp), respectively.
E_Sarbeco Assay:
Forward Primer ACAGGTACGTTAATAGTTAATAGCGT
Reverse Primer ATATTGCAGCAGTACGCACACA
Dual-Labeled Probe ACACTAGCCATCCTTACTGCGCTTCG
Modified RdRp Assay:
Forward Primer AAATGGTCATGTGTGGCGGT
Reverse Primer GTTAAAAACACTATTAGCATAAGCAGTTGT
Dual-Labeled Probe CAGGTGGAACCTCATCAGGAGATGC
The forward primer, reverse primer, and probe are at final concentrations of 8, 8, and 4 μM (E_Sarbeco); and, 10, 10, and 5 μM (Modified RdRp), respectively.
E_Sarbeco Assay:
Forward Primer ACAGGTACGTTAATAGTTAATAGCGT
Reverse Primer ATATTGCAGCAGTACGCACACA
Dual-Labeled Probe ACACTAGCCATCCTTACTGCGCTTCG
Modified RdRp Assay:
Forward Primer AAATGGTCATGTGTGGCGGT
Reverse Primer GTTAAAAACACTATTAGCATAAGCAGTTGT
Dual-Labeled Probe CAGGTGGAACCTCATCAGGAGATGC
Legal Information
FAM is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
Storage Class Code
12 - Non Combustible Liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service