Skip to Content
Merck
All Photos(1)

Key Documents

NCI-H69/LX20

96042332, human lung, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

NCI-H69/LX20, 96042332

biological source

human lung

growth mode

Aggregates in suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian small cell lung carcinoma

Cell Line Description

The drug-resistant cell line NCI-H69/LX20 has been derived by exposure of the parent line, H69 (ECACC Number 91091802) to doxorubicin, also known as adriamycin. These multi drug resistant (MDR) cells have been found to be recognised by various anti-P-glycoprotein antibodies.

Application

Analysis of acquired drug resistance, study of small cell lung carcinoma, MDR studies.

Culture Medium

RPMI 1640 + 2mM Glutamine + 2μg/ml Doxorubicin + 10% Foetal Bovine Serum (FBS). ECACC has found that the following supplement will assist good growth of cells: sodium selenite 10nM, Insulin 250ng/ml, Transferrin 10ug/ml and sodium pyruvate 1mM.

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml; 5% CO2; 37°C. It is recommended to culture the cells without drug upon resuscitation until the first passage. Resuscitate into 9 ml of medium. Immediately after resuscitation perform a centrifugation ste

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service