Skip to Content
Merck
All Photos(1)

Key Documents

MCP21

96030418, Reactivity: Anti-Proteasomes, human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/0

Reactivity

Anti-Proteasomes, human

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Mouse BALB/c spleen

Cell Line Description

Antibodies secreted by the hybridoma line MCP21 react with subunit C3 in human and rabbit proteasomes, both with intact proteasomes and on blots. The hybridoma has been produced by repeated intraperitoneal immunisation of BALB/c mice with 20-50 μg of human placenta proteasomes and subsequent fusion of spleen cells with myeloma Sp2/0.

Immunogen

Human placenta proteasomes

Application

Detection and purification of proteasomes and analysis of subunits

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 5x10,000 and 1x1,000,000 cells/ml; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service