Skip to Content
Merck
All Photos(1)

Key Documents

P1Bb1.1 (DBA/2)

87121102, mouse breast, Mastocytoma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

P1Bb1.1 (DBA/2), 87121102

biological source

mouse breast

growth mode

Suspension

karyotype

Not specified

morphology

Mastocytoma

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse DBA/2 mastocytoma

Cell Line Description

A spontaneous TK- mutant of mastocytoma P815.X2, resistant to BUdR. BUdR is necessary in the growth media to avoid revertants.

Application

Resistance, mutation and tumourigenicity studies

Culture Medium

DMEM + 2mM Glutamine + 100ug/ml Bromo-deoxy Uridine (BUdR) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 8% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service