T47D Cell Line human
85102201, human breast, Epithelial
Synonym(s):
T-47-D Cells, T-47D Cells, T47D:A Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
product name
T47D Cell Line human, 85102201
biological source
human breast
description
Human breast tumor
growth mode
Adherent
karyotype
2n = 46, hypertriploid, modal no. 65
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human breast tumour
Cell Line Description
Established from the pleural effusion of a ductal carcinoma of the breast of a 54-year-old female. The cells carry receptors for a variety of steroids.
Application
T47D cell line human has been used:
- to study the effects of a polysaccharide–protein complex (PPC) isolated from metabolites of gut bacteria Raoultella ornithinolytica from the earthworm Dendrobaena veneta against Candida albicans
- has been cultured and incubated with bone marrow-derived macrophages to study their migration and podosome formation
- to study the anti-proliferative effects of two novel classes of potential antimetabolites on T47D cells
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 11,13
D13S317: 12
D16S539: 10
D5S818: 12
D7S820: 11
THO1: 6
TPOX: 11
vWA: 14
CSF1PO: 11,13
D13S317: 12
D16S539: 10
D5S818: 12
D7S820: 11
THO1: 6
TPOX: 11
vWA: 14
Culture Medium
DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service