Recommended Products
product name
HCA-24, 06061903
biological source
human colon
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
1 of 4
This Item | SMB00161 | SMB00268 | SMB00250 |
---|---|---|---|
≥95% (LC/MS-ELSD) | ≥95% (LC/MS-ELSD) | ≥95% (LC/MS-ELSD) | ≥95% (LC/MS-ELSD) |
metabolomics | metabolomics | metabolomics | metabolomics |
−20°C | −20°C | −20°C | −20°C |
Cell Line Origin
Cell Line Description
Culture Medium
Subculture Routine
Other Notes
Disclaimer
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Need A Sample COA?
This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service