Skip to Content
Merck
All Photos(4)

Key Documents

C8383

Sigma-Aldrich

Chloral hydrate

Synonym(s):

Trichloroacetaldehyde hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCH(OH)2
CAS Number:
Molecular Weight:
165.40
Beilstein:
1698497
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (titration)

Quality Level

form

powder

drug control

USDEA Schedule IV

mp

57 °C (lit.)

SMILES string

OC(O)C(Cl)(Cl)Cl

InChI

1S/C2H3Cl3O2/c3-2(4,5)1(6)7/h1,6-7H

InChI key

RNFNDJAIBTYOQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Chloral hydrate is a sedative/hypnotic.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer Ngolab et al.
Acta neuropathologica communications, 8(1), 123-123 (2020-08-06)
An amendment to this paper has been published and can be accessed via the original article.
Xin Fang et al.
Journal of neurochemistry, 150(4), 441-452 (2019-03-10)
While glucagon-like peptide-1 (GLP-1) was reported to have a positive impact on Parkinson disease, it is extremely short half-life greatly hindered its clinical use. In this study, the mouse strain MG1363-pMG36e-GLP-1 was engineered to continuously express GLP-1 to treat Parkinson
Megan M Monsanto et al.
Nature communications, 11(1), 3955-3955 (2020-08-10)
Cellular therapy to treat heart failure is an ongoing focus of intense research, but progress toward structural and functional recovery remains modest. Engineered augmentation of established cellular effectors overcomes impediments to enhance reparative activity. Such 'next generation' implementation includes delivery
Ya-Fen Jiang-Shieh et al.
Journal of anatomy, 216(3), 407-416 (2010-01-15)
In vivo and in vitro studies have clearly demonstrated that signaling mediated by the interaction of CD200 and its cognate receptor, CD200R, results in an attenuation of inflammatory or autoimmune responses through multiple mechanisms. The present results have shown a
Ya-Chen Ko et al.
Journal of anatomy, 214(1), 183-195 (2009-01-27)
CD200 is a highly glycosylated cell surface protein containing two immunoglobulin superfamily domains in the extracellular region and performs immunosuppressive activities. It is widely distributed in various tissues including the vascular endothelium. We report here the distribution of CD200 in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service