Skip to Content
Merck
All Photos(2)

Key Documents

C7243

Sigma-Aldrich

Chicoric Acid

≥95% (HPLC)

Synonym(s):

2,3-Dicaffeoyl-L-tartaric acid, Cichoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18O12
CAS Number:
Molecular Weight:
474.37
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

OC1=CC=C(/C=C/C(O[C@@H](C(O)=O)[C@H](C(O)=O)OC(/C=C/C2=CC(O)=C(O)C=C2)=O)=O)C=C1O

InChI

1S/C22H18O12/c23-13-5-1-11(9-15(13)25)3-7-17(27)33-19(21(29)30)20(22(31)32)34-18(28)8-4-12-2-6-14(24)16(26)10-12/h1-10,19-20,23-26H,(H,29,30)(H,31,32)/b7-3+,8-4+/t19-,20-/m1/s1

InChI key

YDDGKXBLOXEEMN-IABMMNSOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Pharmacokinetics and antioxidant activity of dihydrocaffeic acid grafted chitosan nanomicelles loaded with chicoric acid in broilers.: This study investigates the pharmacokinetics and antioxidant properties of chicoric acid-loaded nanomicelles in broilers, highlighting its potential for improving poultry health (Ren et al., 2024).

  • Screening of the effective sites of Cichorium glandulosum against hyperuricemia combined with hyperlipidemia and its network pharmacology analysis.: The research identifies effective components in Cichorium glandulosum, including chicoric acid, for treating hyperuricemia and hyperlipidemia using network pharmacology methods (Yang et al., 2024).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Santander et al.
Journal of the science of food and agriculture, 100(4), 1577-1587 (2019-11-27)
Arbuscular mycorrhizal (AM) fungi establish symbioses with most agricultural plants and improves growth under soil stress conditions. The present study aimed to evaluate the functional contribution of 2 AM fungal inocula (a native consortium isolated from saline soils of the Atacama

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service