Skip to Content
Merck
All Photos(1)

Key Documents

C6146

Sigma-Aldrich

2-Butenoyl coenzyme A lithium salt

≥90% (HPLC)

Synonym(s):

2-Butenoyl CoA lithium salt, Crotonoyl Co A lithium salt, Crotonoyl coenzyme A lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H40N7O17P3S
CAS Number:
Molecular Weight:
835.61
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Quality Level

Assay

≥90% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

[Li].C\C=C\C(=O)SCCNC(=O)CCNC(=O)C(O)C(C)(C)COP(O)(=O)OP(O)(=O)OCC1OC(C(O)C1OP(O)(O)=O)n2cnc3c(N)ncnc23

InChI

1S/C25H40N7O17P3S.Li.H/c1-4-5-16(34)53-9-8-27-15(33)6-7-28-23(37)20(36)25(2,3)11-46-52(43,44)49-51(41,42)45-10-14-19(48-50(38,39)40)18(35)24(47-14)32-13-31-17-21(26)29-12-30-22(17)32;;/h4-5,12-14,18-20,24,35-36H,6-11H2,1-3H3,(H,27,33)(H,28,37)(H,41,42)(H,43,44)(H2,26,29,30)(H2,38,39,40);;/b5-4+;;

InChI key

OXXVXWRNRODUOD-SFKRKKMESA-N

Related Categories

Application

Crotonoyl coenzyme A (2-Butenoyl coenzyme A) may be used as a substrate of enzymes such as Plasmodium falciparum enoyl-ACP reductase and other enoyl-CoA reductases. Crotonoyl –CoA may be used as a substrate analogue for kinetic studies on β-hydroxyacyl-acyl carrier protein (ACP) dehydratase (FabZ).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weizhi Liu et al.
Biochemical and biophysical research communications, 333(4), 1078-1086 (2005-06-22)
Helicobacter pylori is a gram-negative pathogenic bacterium that causes peptic ulcer disease and gastric cancer, and studies of the related potent enzymes associated with this bacterium are urgent for the discovery of novel drug targets. In bacteria, beta-hydroxyacyl-acyl carrier protein
Mili Kapoor et al.
The Biochemical journal, 381(Pt 3), 725-733 (2004-05-06)
The binding of enoyl-ACP (acyl-carrier protein) reductase from Plasmodium falciparum (PfENR) with its substrates and inhibitors has been analysed by SPR (surface plasmon resonance). The binding of the substrate analogue crotonoyl-CoA and coenzyme NADH to PfENR was monitored in real
Sara Tucci et al.
FEBS letters, 581(8), 1561-1566 (2007-03-27)
An NADH-dependent trans-2-enoyl-CoA reductase (EC1.1.1.36) from the Gram negative spirochete Treponema denticola was identified, expressed and biochemically characterized. The recombinant protein is a monomeric enzyme with a molecular mass of 44 kDa with a specific activity of 43+/-4.8 U/mg (micromol
Neha Kapoor et al.
IUBMB life, 61(11), 1083-1091 (2009-10-28)
A structure-based approach has been adopted to develop 2'-substituted analogs of triclosan. The Cl at position 2' in ring B of triclosan was chemically substituted with other functional groups like NH(2), NO(2) and their inhibitory potencies against PfENR were determined.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service